Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RAR gamma-1 Antibodies

          Invitrogen

          RAR gamma-1 Recombinant Rabbit Monoclonal Antibody (PSH07-19)

          Advanced Verification
          View all (11) RAR gamma-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RAR gamma-1 Recombinant Rabbit Monoclonal Antibody (PSH07-19)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          • Advanced Verification (3)
          RAR gamma-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RAR gamma-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAR gamma-1 Antibody (MA5-60185) in IHC (P)

          Immunohistochemical analysis of RAR gamma-1 on formalin-fixed paraffin-embedded mouse skin tissue. The section was pre-treated using heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0) for 20 minutes. The tissue was blocked in 1% BSA for 20 minutes at room temperature, then probed with RAR gamma-1 Recombinant Rabbit Monoclonal Antibody (PSH07-19) (Product # MA5-60185) at 1:1,000 dilution for 1 ho... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAR gamma-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAR gamma-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAR gamma-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RAR gamma-1 Antibody in Western Blot (WB)
          RAR gamma-1 Antibody in Western Blot (WB)
          RAR gamma-1 Antibody in Western Blot (WB)
          RAR gamma-1 Antibody in Western Blot (WB)
          RAR gamma-1 Antibody in Western Blot (WB)
          RAR gamma-1 Antibody
          RAR gamma-1 Antibody
          RAR gamma-1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RAR gamma-1 Recombinant Rabbit Monoclonal Antibody (PSH07-19)

          Product Details

          MA5-60185

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,00-1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000-1:5,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH07-19

          Immunogen

          Recombinant protein within human RARG aa 155-454.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.051 mg/mL

          Amount

          105.1 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          RRID

          AB_3683087

          Product Specific Information

          Positive Control: SK-Br-3 cell lysate, T-47D cell lysate, A375 cell lysate, HeLa cell lysate, MEF cell lysate, Mouse skin tissue lysate, Rat skin tissue lysate, human skin tissue, mouse skin tissue, rat skin tissue.

          Tissue Specificity: Tissue enhanced (esophagus, skin).

          Subcellular Location: Nucleus, Cytoplasm.

          Sequence Similarities: 100% Mouse/Rat.

          Predicted band size: 50 kDa.

          Target Information

          Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: mRAR&agr; homolog; nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR Gamma-1; RAR-gamma; retinoic acid; retinoic acid receptor; retinoic acid receptor gamma; retinoic acid receptor gamma 1; retinoic acid receptor gamma 2; retinoic acid receptor gamma 3; retinoic acid receptor gamma 4; unnamed protein product

          View more View less

          Gene Aliases: NR1B3; RAR-gamma; RARC; RARD; RARgamma; RARgamma2

          View more View less

          UniProt ID: (Human) P13631, (Mouse) P18911

          View more View less

          Entrez Gene ID: (Human) 5916, (Mouse) 19411, (Rat) 685072

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding sequence-specific DNA binding retinoid X receptor binding sequence-specific double-stranded DNA binding RNA polymerase II regulatory region sequence-specific DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neural tube closure chondrocyte development glandular epithelial cell development retinal pigment epithelium development growth plate cartilage development growth plate cartilage chondrocyte growth regulation of transcription, DNA-templated positive regulation of cell proliferation negative regulation of cell proliferation anterior/posterior pattern specification regulation of gene expression positive regulation of gene expression cell differentiation embryonic camera-type eye development regulation of myelination negative regulation of chondrocyte differentiation response to retinoic acid embryonic hindlimb morphogenesis multicellular organism growth camera-type eye development positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of programmed cell death negative regulation of cell differentiation regulation of myeloid cell differentiation positive regulation of transcription from RNA polymerase II promoter embryonic eye morphogenesis retinoic acid receptor signaling pathway embryonic organ development embryonic morphogenesis reproductive structure development gland development retina development in camera-type eye limb development face development bone development bone morphogenesis epithelium development trachea cartilage development prostate gland epithelium morphogenesis negative regulation of cartilage development Harderian gland development cellular response to retinoic acid cellular response to leukemia inhibitory factor regulation of cell size canonical Wnt signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline