Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-12 p35 Antibodies

          Invitrogen

          IL-12 p35 Monoclonal Antibody (116219)

          View all (41) IL-12 p35 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-12 p35 Monoclonal Antibody (116219)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA5-60931

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Flow Cytometry (Flow)

          2.5 µg/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Pig

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          116219

          Immunogen

          Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p35/p40 heterodimer, p40 Ile23-Asn324, p35 Arg26-Ser222

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.5 mg/mL

          Amount

          500 µg

          Purification

          Protein A/G

          Storage buffer

          PBS with trehalose

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3701543

          Product Specific Information

          Reconstitute at 0.5 mg/mL in sterile PBS.

          Target Information

          Interleukin 12A is a novel signaling cytokine belonging to the Interleukin-6 protein superfamily. Interleukin-12 alpha chain precursor (IL-12A) is a 35-kD subunit of disulfide-linked heterodimer of cytokine IL-12 and has a broad array of biological activities. Reports suggest that IL-12A can act as a growth factor for activated T and NK cells and enhance the lytic activity of NK/lymphokine-activated Killer cells. IL-12A stimulates the production of IFN-gamma by resting PBMC as well as T-cell-independent induction of interferon (IFN)-gamma, and helps in the differentiation of both Th1 and Th2 cells. It is also involved in positive regulation of tyrosine phosphorylation of Stat4 protein. The signaling process of IL-12A in innate immunity requires nitric oxide synthase 2A. Along with EBI3, IL-12A forms a novel heterodimeric cytokine, designated as interleukin-35 (IL-35), that may be specifically produced by T reg cells and is required for maximal suppressive activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CLMF p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12A; IL12p35; ILN; Interleukin; Interleukin 12; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 p35 subunit; Interleukin12

          View more View less

          UniProt ID: (Pig) Q29053

          View more View less

          Entrez Gene ID: (Pig) 397053

          View more View less

          Function(s)
          cytokine activity interleukin-12 receptor binding growth factor activity interleukin-12 beta subunit binding interleukin-27 binding protein heterodimerization activity
          Process(es)
          positive regulation of T cell mediated cytotoxicity positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target cell cycle arrest cell proliferation response to UV-B cell migration negative regulation of interleukin-17 production positive regulation of interferon-gamma production positive regulation of natural killer cell activation positive regulation of smooth muscle cell apoptotic process T-helper 1 cell activation T-helper 1 cell cytokine production positive regulation of T cell proliferation positive regulation of tyrosine phosphorylation of STAT protein defense response to protozoan positive regulation of T cell differentiation negative regulation of smooth muscle cell proliferation negative regulation of protein secretion defense response to Gram-positive bacterium positive regulation of NK T cell activation cellular response to lipopolysaccharide extrinsic apoptotic signaling pathway cellular response to virus negative regulation of vascular endothelial growth factor signaling pathway negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis positive regulation of dendritic cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline