Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
CNP (2',3'-Cyclic Nucleotide 3' Phosphodiesterase) is a Protein Coding gene. Diseases associated with CNP include Leukodystrophy, Hypomyelinating, 20 and Myopia 2, Autosomal Dominant. Among its related pathways are MECP2 and associated Rett syndrome and Glial cell differentiation. Gene Ontology (GO) annotations related to this gene include RNA binding and cyclic nucleotide binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2'',3''-cyclic nucleotide 3'' phosphodiesterase; 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase (EC 3.1.4.37); CNP1; CNPase; HLD20; unnamed protein product
Gene Aliases: CN37; CNP1; HLD20
UniProt ID: (Human) P09543
Entrez Gene ID: (Human) 1267
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support