Murine Dendritic Cell Cytokine Package, PeproTech®
Gibco™
Murine Dendritic Cell Cytokine Package, PeproTech®
The Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
Have Questions?
Catalog NumberQuantity
MDC1 Ea.
Catalog number MDC
Price (USD)
774.65
Online exclusive
1,032.00
Save 257.35
Each
Quantity:
1 Ea.
Price (USD)
774.65
Online exclusive
1,032.00
Save 257.35
Each
The Murine Dendritic Cell Cytokine Package contains IL-4 and GM-CSF, key components required for the culturing of murine dendritic cells. Recombinant murine IL-4 is an essential cytokine used in the differentiation and maturation of murine dendritic cell in vitro. The IL-4 protein, specifically recombinant IL-4, promotes the development of dendritic cells from precursor cells such as monocytes. Murine IL-4 acts synergistically with other factors, including GM-CSF (granulocyte-macrophage colony stimulating factor), to drive the differentiation process. Typically, monocytes are cultured in the presence of recombinant IL-4 and GM-CSF to yield immature dendritic cells, which can then be futher matured by additional stimuli.
The package contains the following two vials:
  • 100 μg Murine IL-4 (Cat. No. 214-14-100UG)
  • 100 μg Murine GM-CSF (Cat. No. 315-03-100UG)
IL-4 is a pleiotropic cytokine that regulates diverse T- and B-cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
Specifications
For Use With (Application)Functional Assay
FormulationLyophilized
Quantity1 Ea.
SpeciesMouse
Product TypeDendritic Cell Cytokine Package
Unit SizeEach
Frequently asked questions (FAQs)
Which cytokines show cross-species activity?
With a few exceptions, most human cytokines are active on mouse cells. Many mouse cytokines are active on human cells, but may show lower specific activity than the corresponding human cytokine. The interferons, GM-CSF, IL-3, and IL-4 are known to be species-specific with very little, if any, activity on non-homologous cells. In contrast, the FGFs and neurotrophins are very highly conserved and show excellent activity on cells of other animal species.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.