Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • hCG Antibodies

          Invitrogen

          hCG Monoclonal Antibody (3015)

          View all (15) hCG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite hCG Monoclonal Antibody (3015)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          hCG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          hCG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          hCG Antibody (MIH9805) in IHC (P)

          Immunohistochemistry analysis of hCG was performed on human placenta tissue and is showing positive staining in extra cellular spaces. To expose target proteins, antigen retrieval was performed by microwaving tissues for 20 minutes in 10mM sodium citrate buffer (pH 6.0). Tissue slides were probed with hCG monoclonal antibody (Product # MIH9805) at a dilution of 1:20, overnight at 4C in a humidified chamber ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          hCG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hCG Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MIH9805

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250-1:500
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Radioimmune Assays (RIA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3015

          Immunogen

          Human Chorionic Gonadotropin

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          5 mg/mL

          Amount

          1 mg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11152013

          Product Specific Information

          The observed molecular weight of alpha-hCG is approximately 20-22 kD. The molecular weight is higher than the predicted molecular weight due to post translational modification.

          MIH9805 was formerly sold as a Seradyn product.

          Target Information

          Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregnancy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: anterior pituitary glycoprotein hormones common subunit alpha; CGA; choriogonadotropin alpha chain; chorionic gonadotrophin chain beta; chorionic gonadotrophin subunit alpha; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta 5 subunit; chorionic gonadotropin beta 8 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit 3; chorionic gonadotropin beta subunit 5; chorionic gonadotropin beta subunit 8; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, alpha polypeptide; chorionic gonadotropin, beta polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSH-alpha; human chorionic gonadotropin; LSH-alpha; luteinizing hormone alpha chain; lutropin alpha chain; thyroid-stimulating hormone alpha chain; thyrotropin alpha chain; TSH-alpha; unnamed protein product

          View more View less

          Gene Aliases: CG-ALPHA; CG-beta; CGB; FSHA; GPA1; GPHa; GPHA1; HCG; LHA; TSHA

          View more View less

          UniProt ID: (Human) P01215, (Human) P0DN86

          View more View less

          Entrez Gene ID: (Human) 1081, (Human) 1082, (Human) 93659, (Human) 94115

          View more View less

          Function(s)
          hormone activity protein binding follicle-stimulating hormone activity receptor binding
          Process(es)
          apoptotic process signal transduction G-protein coupled receptor signaling pathway cell-cell signaling female gamete generation hormone-mediated signaling pathway MAPK cascade ovarian follicle development spermatogenesis positive regulation of cell proliferation regulation of receptor activity positive regulation of steroid biosynthetic process positive regulation of cell migration insulin secretion involved in cellular response to glucose stimulus follicle-stimulating hormone signaling pathway positive regulation of transcription from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline