Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-12 p70 Antibodies

          Invitrogen

          IL-12 p70 Monoclonal Antibody (C17-8), Biotin

          19 References
          View all (25) IL-12 p70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-12 p70 Monoclonal Antibody (C17-8), Biotin

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}

          Product Details

          MM121B

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          View 14 publications 14 publications

          Neutralization (Neu)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C17-8

          Immunogen

          Recombinant mouse IL-12

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          250 µg

          Purification

          Protein G

          Storage buffer

          PBS with 4% BSA

          Contains

          proprietary Preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_223600

          Product Specific Information

          MM121B targets IL-12 in ELISA, and WB applications and shows reactivity with mouse samples.



          The MM121B immunogen is recombinant mouse IL-12.



          MM121B detects IL-12 which has a predicted molecular weight of approximately 22 kDa.



          The MM121B mouse IL12 antibody (biotinylated conjugate of clone C17-8) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody MM120 (clone 9A5). Typical dilutions for sandwich ELISA include 1 µg/mL for coating and 0.125 - 0.25 µg/mL for detection.



          Biotinylated antibody MM121B (clone C17-8) and antibody MM120 (clone 9A5) have successfully been used in combination with recombinant mouse IL12p70 protein SMIL12 in ELISA applications.



          This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified.

          Target Information

          IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 20C2; CLMF p35; CLMF p40; cytotoxic lymphocyte maturation factor 35 kDa subunit; cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; IL-12 p35 subunit; IL-12 p40 subunit; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-23 subunit p40; IL12A/B; IL12AB; ILN; Interleukin; interleukin 12a p35 subunit; interleukin-12 p35 subunit; interleukin-12 p40 subunit; Interleukin-12 p70; MGC151228; MGC151232; unnamed protein product

          View more View less

          Gene Aliases: Il-12a; Il-12b; IL-12p35; Il-12p40; Il12p40; Ll12a; p35; p40

          View more View less

          UniProt ID: (Mouse) P43431, (Mouse) P43432

          View more View less

          Entrez Gene ID: (Mouse) 16159, (Mouse) 16160

          View more View less

          Function(s)
          cytokine activity interleukin-12 receptor binding protein binding growth factor activity interleukin-12 beta subunit binding interleukin-27 binding protein heterodimerization activity cytokine receptor activity cytokine receptor binding interleukin-12 alpha subunit binding identical protein binding macromolecular complex binding interleukin-23 receptor binding
          Process(es)
          positive regulation of leukocyte mediated cytotoxicity positive regulation of T cell mediated cytotoxicity positive regulation of defense response to virus by host natural killer cell activation involved in immune response positive regulation of lymphocyte mediated immunity positive regulation of T-helper 1 type immune response positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target negative regulation of inflammatory response to antigenic stimulus cell surface receptor signaling pathway JAK-STAT cascade cell proliferation response to UV-B cell migration natural killer cell activation negative regulation of interleukin-10 production negative regulation of interleukin-17 production positive regulation of granulocyte macrophage colony-stimulating factor production positive regulation of interferon-gamma production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-17 production positive regulation of tumor necrosis factor production positive regulation of natural killer cell activation positive regulation of natural killer cell proliferation positive regulation of mononuclear cell proliferation positive regulation of smooth muscle cell apoptotic process interleukin-12-mediated signaling pathway interleukin-23-mediated signaling pathway T-helper cell differentiation positive regulation of T cell proliferation positive regulation of activated T cell proliferation defense response to protozoan T-helper 1 cell differentiation positive regulation of osteoclast differentiation positive regulation of cell adhesion positive regulation of JAK-STAT cascade negative regulation of smooth muscle cell proliferation positive regulation of lymphocyte proliferation negative regulation of protein secretion regulation of inflammatory response defense response to Gram-negative bacterium positive regulation of NK T cell activation positive regulation of NK T cell proliferation defense response to virus cellular response to lipopolysaccharide cellular response to interferon-gamma negative regulation of vascular endothelial growth factor signaling pathway negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis positive regulation of T-helper 17 type immune response immune response response to virus response to lipopolysaccharide T-helper 1 cell activation positive regulation of tyrosine phosphorylation of STAT protein positive regulation of T cell differentiation positive regulation of natural killer cell mediated cytotoxicity defense response to Gram-positive bacterium extrinsic apoptotic signaling pathway cellular response to virus positive regulation of dendritic cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0