Motorized Tilt & Rotation Sample Holder
Thermo Scientific™
Motorized Tilt & Rotation Sample Holder
Revealing the hidden treasures of all features on your sample is now possible with the Motorized Tilt & Rotation Sample Holder. Samples can have lines and holes, or have multi-layer structures. The new Motorized Tilt & Rotation Sample Holder allows analysis of the sample from all visible sides and enables a unique 3D image of your sample.
Have Questions?
Número de catálogo MOTILTROTSAMPLHOLD
Precio (USD)
-
Revealing the hidden treasures of all features on your sample is now possible with the Thermo Scientific™ Motorized Tilt & Rotation Sample Holder. Samples can have lines and holes, or have multi-layer structures. The new Motorized Tilt & Rotation Sample Holder allows analysis of the sample from all visible sides and enables a unique 3D image of your sample.
The Motorized Tilt & Rotation Sample holder is a so-called smart sample holder that does not have any cables attached. The sample holder interface board automatically identifies the sample holder type and will directly transfer tasks to the holder.
Key Specifications
  • Tilt range -10° to +45°
  • Continuous 360° compucentric rotation
  • Controlled by dedicated Motion Control ProSuite application
  • Tilt adapted focus
  • Align X and Y
Especificaciones
Unit SizeEach