VetMAX™ M. bovis Kit
Applied Biosystems™

VetMAX™ M. bovis Kit

Note: The name of this product has changed. It was previously called LSI VetMAX MYCOPLASMA BOVIS. Same as TaqVet MycoplasmaRead more
Have Questions?
Catalog NumberQuantity
MPBO5050 tests
Catalog number MPBO50
Price (USD)
-
Quantity:
50 tests
Note: The name of this product has changed. It was previously called LSI VetMAX MYCOPLASMA BOVIS.

Same as TaqVet Mycoplasma bovis Real-Time PCR Kit (MPBO)

Real-time PCR enables sensitive and specific detection of pathogen nucleic acid in animal samples, allowing for reliable and rapid screening and detection of infected animals.

The LSI VetMAX™ kits include:
• 1 ready-to-use mix that includes a set of oligonucleotides for detection of the target gene OR these separate components that will need to be mixed together: a set of oligonucleotides for the detection of the target gene + a TaqMan™ real-time PCR master mix + an RT enzyme (only for RNA kits)
• 1 external positive control of the same nature as the target (inactivated bacteria or virus). Use as a quality control for PCR and/or extraction yield.
• A quality control certificate

For veterinary use only.

Regulatory requirements vary by country; products may not be available in your geographic area.

WARNING! BIOHAZARD. Biological samples such as tissues, body fluids, infectious agents, and blood of humans and other animals have the potential to transmit infectious diseases. All work should be conducted in properly equipped facilities using the appropriate safety equipment (for example, physical containment devices). Safety equipment also may include items for personal protection, such as gloves, coats, gowns, shoe covers, boots, respirators, face shields, safety glasses, or goggles. Individuals should be trained according to applicable regulatory and company/institution requirements before working with potentially biohazardous materials. Follow all applicable local, state/provincial, and/or national regulations. The following references provide general guidelines when handling biological samples in laboratory environment.

1. U.S. Department of Health and Human Services, Biosafety in Microbiological and Biomedical Laboratories (BMBL), 5th Edition, HHS Publication No. (CDC) 21-1112, Revised December 2009; found at: www.cdc.gov/biosafety/publications/bmbl5/BMBL.pdf.

2. World Health Organization, Laboratory Biosafety Manual, 3rd Edition, WHO/CDS/CSR/LYO/2004.11; found at: www.who.int/csr/resources/publications/biosafety/Biosafety7.pdf.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Specifications
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity50 tests
Sample TypeAny Organ Type, Bronchoalveolar Lavage (BAL), Synovial Fluid, Milk
SpeciesCattle/Bovine
TargetpolC
Target Organism ClassM. bovis
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceLow
Unit SizeEach

Frequently asked questions (FAQs)

What kind of milk samples can be tested with the VetMAX MastiType kits?

All VetMAX MastiType kits may be used with milk, bulk milk, frozen milk, or preserved milk (e.g., with bronopol).

What qPCR instrument can I use to run the VetMAX MastiType kits?

Either the Applied Biosystems 7500/7500 Fast Real-Time PCR System or the QuantStudio 5 Real-Time PCR System may be used to run the VetMAX MastiType kits.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Which sample prep kit do you recommend upstream of the VetMAX MastiType kits?

We recommend using the MagMAX CORE Nucleic Acid Purification Kit + MagMAX CORE Mastitis & Panbacteria Module (Cat. No. A40289) with the VetMAX MastiType kits.

Which organisms can be detected by each VetMAX MastiType kit?

The VetMax MastiType Micro4 Kit detects Mycoplasma bovis, Staphylococcus aureus, Streptococcus agalactiae, and Streptococcus uberis.

The VetMax MastiType Myco8 Kit detects Mycoplasma spp., Mycoplasma alkalescens, Mycoplasma bovis, Mycoplasma bovigenitalium, Mycoplasma canadense, Mycoplasma californicum, Staphylococcus aureus, and Streptococcus agalactiae.

The VetMax MastiType Multi Kit detects Staphylococcus aureus, Staphylococcus spp. (including all major coagulase-negative staphylococci), Streptococcus agalactiae, Streptococcus dysgalactiae, Streptococcus uberis, Escherichia coli, Enterococcus spp. (incl. E. faecalis and E. faecium), Klebsiella oxytoca and/or Klebsiella pneumoniae, Serratia marcescens, Corynebacterium bovis, Trueperella pyogenes and/or Peptoniphilus indolicus, Staphylococcal beta-lactamase gene (penicillin-resistance gene), Mycoplasma bovis, Mycoplasma spp., Yeast, and Prototheca spp.