Can Lipofectamine LTX be used for transfecting primary cells?
We have tested four primary cell lines: HuVec, NHFF (Normal human foreskin fibroblast), MJ90 and NDHF. We have observed transfection efficiency in the range of 5-25%. We suggest optimizing transfection efficiency for your primary cell line by varying DNA and lipid ratio.
If you are using any other primary cell line, please follow guidelines suggested in the product insert. Alternatively, Neon electroporation system and viral delivery methods are recommended.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.