VetMAX™ M. tuberculosis Complex PCR Kit - FAQs
VetMAX™ M. tuberculosis Complex PCR Kit - FAQs
View additional product information for VetMAX™ M. tuberculosis Complex PCR Kit - FAQs (MTBC)
1 product FAQs found
What are the probes included in the VetMAX M. tuberculosis Complex PCR Kit?
The TaqMan assay for TUB is labeled with FAM and quenched by NFQ (non-fluorescent quencher). The TaqMan assay for IPC is labeled with VIC and quenched by NFQ (non-fluorescent quencher).