Slideport™ Slide Inserts
Thermo Scientific™
Slideport™ Slide Inserts
The Thermo Scientific™ Slideport™ Slide Insert holds microscope slides in a frame that fits Thermo Scientific™ High Content Instruments. SlidesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
N01011133 Pk.
N01011111 pack
N01011155 pack
Catalog number N0101113
Price (USD)
430.00
Each
Quantity:
3 Pk.
Price (USD)
430.00
Each
The Thermo Scientific™ Slideport™ Slide Insert holds microscope slides in a frame that fits Thermo Scientific™ High Content Instruments. Slides can be imaged using many objective lenses due to two available skirt heights. The Slideport Slide Insert has been tested to image a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays and beads. Contents: five Slideport Slide Inserts.
Features of Slideport Slide Inserts:
• Two skirt heights available to accommodate a variety of readers and objectives
• Any microscope or plate reader that accepts standard microplates will also accept the Slideport Slide Insert
• Image a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays, and beads
• Easily stack slides for automated imaging with or without lids
• Slideport Slide Insert can be utilized at a variety of temperatures from 4°to 37°C, allowing for flexibility of storage and imaging conditions
Compatible with the Thermo Scientific High Content Instruments
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)CellInsight CX5 HCS Platform, ArrayScan HCA Reader
Holds4 slides per insert
Quantity3 Pk.
Product LineSlideport
TypeSlide Insert
Unit SizeEach