Product Image
Thermo Scientific™
ArrayScan™ 7-Color LED Module for ArrayScan™ HCA Readers
The Thermo Scientific™ ArrayScan™ 7-Color LED Module for ArrayScan™ HCA Readers (600-800 series) is a seven color, direct coupled, LightRead more
Have Questions?
Catalog NumberQuantity
N010LED71 each
Catalog number N010LED7
Price (USD)
-
Quantity:
1 each
The Thermo Scientific™ ArrayScan™ 7-Color LED Module for ArrayScan™ HCA Readers (600-800 series) is a seven color, direct coupled, Light Emitting Diode (LED) light source designed for high performance and ultra-reliability to excite fluorochromes that are blue, cyan, green, yellow, orange-red, scarlet, far red, and near IR.
Features of the ArrayScan 7-Color LED Module for ArrayScan HCA Readers:
• Spectral flexibility—spanning the visible spectrum from UV through near-IR, the ArrayScan 7-Color LED Module can use the available filter combinations to provide for a multitude of excitation and emission combinations
• Spectral separation—avoid bleed through between channels by using excitation and emission wavelengths that have bands separated enough to prevent spectral overlap
Includes:
• To be used with Thermo Scientific™ HCS Studio™ Cell Analysis Software (version 2.0 or greater) and minimum computer specifications
• Filters and emission filter wheel
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)ArrayScan HCA Readers (600-800 Series)
Operating SystemRequires Windows 7 supported minimum computer specifications and HCS Studio from supplied from Thermo Fisher Scientific
Quantity1 each
Software CompatibilityThermo Scientific™ HCS Studio Cell Analysis Software
Product LineArrayScan
Unit Size1 each