HRP-Conjugated Streptavidin - FAQs
HRP-Conjugated Streptavidin - FAQs
View additional product information for HRP-Conjugated Streptavidin - FAQs (N100)
1 product FAQs found
In which solvent is the HRP-Conjugated Streptavidin (Cat. No. N100) dissolved?
The HRP-Conjugated Streptavidin (Cat. No. N100) is dissolved in an aqueous solution of 0.99% BSA and 0.1% ProCelin preservative.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.