Barcode Reader for KingFisher™ Duo Prime - FAQs
Barcode Reader for KingFisher™ Duo Prime - FAQs
View additional product information for Barcode Reader for KingFisher™ Duo Prime - FAQs (N16640)
1 product FAQs found
Are the Matrix 500 microliter ScrewTop Tubes (Cat. No. 3743AMB) compatible with the Kingfisher Duo Prime barcode reader?
Yes, the KingFisher Duo Prime barcode reader is able to read the 2D barcode on the Matrix 500 µL ScrewTop Tubes. The barcode reader can generate an error if the tubes are in the tube rack and the barcode position is not able to be scanned. If this occurs, we recommend scanning each tube individually.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.