Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip - FAQs
Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip - FAQs
View additional product information for Multidrop Combi SMART+ Small Tube Cassette, Plastic Tip - FAQs (N22700)
1 product FAQs found
What are the tubing lengths of the regular length and extended length cassettes for the Multidrop Combi Reagent Dispensers?
Regular length tubing (small and standard) cassette length is 40 cm long.
Extended length tubing (small and standard) cassette length is 200 cm long.
Note: You can cut the extended length tubing to the desired length but we recommend checking the calibration on the cassette, once modified, to ensure it is providing accurate results.