T7 Promoter Primer - FAQs
T7 Promoter Primer - FAQs
View additional product information for T7 Promoter Primer - FAQs (N56002)
2 product FAQs found
How is the T7 Promoter Primer (Cat. No. N56002) supplied?
It is a desalted primer supplied lyophilized in 10 mM Tris-HCl, pH 7.5, 1 mM EDTA
What is the sequence of the T7 Promoter Primer (Cat. No. N56002)?
Here is the sequence:
TAATACGACTCACTATAGGG