Nitro blue Tetrazolium Chloride (NBT)
Invitrogen™
Nitro blue Tetrazolium Chloride (NBT)
Use to assess cell viability as function of redox potential
Have Questions?
Catalog NumberQuantity
N64951 g
Catalog number N6495
Price (MXN)
-
Quantity:
1 g
Nitro blue tetrazolium chloride (NBT) can be used to assess cell viability as a function of redox potential. Actively respiring cells convert the water-soluble NBT to a deep blue-colored precipitate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 g
Shipping ConditionRoom Temperature
SubstrateOxidase Substrate
Absorbance Wavelength256 nm
Detection MethodAbsorbance
Substrate PropertiesChemical Substrate
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.
Citations & References (166)
Citations & References
Abstract
Endogenous endothelial cell nitric-oxide synthase modulates apoptosis in cultured breast cancer cells and is transcriptionally regulated by p53.
Authors:Mortensen K,Skouv J,Hougaard DM,Larsson LI
Journal:The Journal of biological chemistry
PubMed ID:10608825
In situ polymerase chain reaction amplification of HIV-1 DNA in brain tissue.
Authors:Strappe PM, Wang TH, McKenzie CA, Lowrie S, Simmonds P, Bell JE
Journal:J Virol Methods
PubMed ID:9562406
A direct in situ polymerase chain reaction (IS-PCR) assay is described for the detection of HIV-1 proviral DNA in formalin fixed paraffin embedded brain tissue. Biotin-16-dUTP is incorporated during the PCR process and microwave pretreatment of tissue sections ensures that no non-specific incorporation into damaged or nicked genomic DNA occurs. ... More
Study of chronic granulomatous disease by a nitroblue tetrazolium densitometric kinetic test: a new research method.
Authors:Iacobini M, Torre A, Palumbo G, Del Principe D, Digilio G, Balducci L
Journal:Clin Chim Acta
PubMed ID:8149637
Ectopic expression of Drosophila ELAV and human HuD in Drosophila wing disc cells reveals functional distinctions and similarities.
Authors:Toba G, Qui J, Koushika SP, White K
Journal:J Cell Sci
PubMed ID:12006625
'Drosophila ELAV and human HuD are two neuronal RNA binding proteins that show remarkable sequence homology, yet differ in their respective documented roles in post-transcriptional regulation. ELAV regulates neural-specific alternative splicing of specific transcripts, and HuD stabilizes specific mRNAs that are otherwise unstable due to AU-rich elements (AREs) in their ... More
Regulation of chemotactic and proadhesive responses to chemoattractant receptors by RGS (regulator of G-protein signaling) family members.
Authors:Bowman EP, Campbell JJ, Druey KM, Scheschonka A, Kehrl JH, Butcher EC
Journal:J Biol Chem
PubMed ID:9774420
'Serpentine Galphai-linked receptors support rapid adhesion and directed migration of leukocytes and other cell types. The intracellular mechanisms mediating and regulating chemoattractant-directed adhesion and locomotion are only now beginning to be explored. RGS (for regulator of G-protein signaling) proteins are a recently described family that regulate Galphai-stimulated pathways by acting ... More