MicroAmp™ 8-Cap Strip, clear, 300 Strips - FAQs
MicroAmp™ 8-Cap Strip, clear, 300 Strips - FAQs
View additional product information for MicroAmp™ 8-Cap Strip, clear - FAQs (N8010535, N8011535)
1 product FAQs found
Can I use caps on the 96-well plates on the QuantStudio 12K Flex system?
Yes. We recommend that you use Optical Flat caps, such as Cat. No. N8010535. Use of domed caps may damage the heated cover.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.