TaqMan™ Reverse Transcription Reagents, 200 reactions - FAQs
TaqMan™ Reverse Transcription Reagents, 200 reactions - FAQs
View additional product information for TaqMan™ Reverse Transcription Reagents - FAQs (4304134, N8080234)
1 product FAQs found
Can the RT buffer for TaqMan Reverse Transcription Reagents kit (Cat. No. N8080234) be purchased separately?
Yes, it is the same as the GeneAmp 10X PCR Buffer II & MgCl2 (Cat. No. N8080010).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.