NanoDrop™ 2000/2000c Spectrophotometers
Thermo Scientific™

NanoDrop™ 2000/2000c Spectrophotometers

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop Ultra Microvolume UV-Vis Spectrophotometer.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
ND-2000CSpectrophotometern.a.
ND-2000Microvolume Spectrophotometern.a.
ND2000CLAPTOPUV-Vis Spec + Laptop ComputerLaptop Computer
ND2000LAPTOPSpectrophotometerLaptop Computer
Catalog number ND-2000C
Price (USD)
-
Type:
Spectrophotometer
Includes:
n.a.

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop Ultra Microvolume UV-Vis Spectrophotometer.

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop Ultra Microvolume UV-Vis Spectrophotometer.

US Patents 6,628,382 and 6,809,826.
Specifications
Absorbance Accuracy3% (at 0.74 Abs at 350 nm)
Absorbance RangePedestal: 0 to 300 A Cuvette: 0 to 1.5 A
Accuracy0.002
Certifications/ComplianceUL/CSA and CE
CompatibilityMicrosoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
ConnectionsUSB
Depth (Metric)20 cm
DescriptionMicrovolume Spectrophotometer, with cuvette capability
Detection LimitsPedestal: 2 ng/μL (dsDNA), 0.10 mg/mL (BSA) Cuvette: 0.4 ng/μL (dsDNA), 0.01 mg/mL (BSA)
Detection Range2 to 15,000n g/μL (dsDNA) , 0.10 to 400 mg/mL (BSA)
Footprint14 x 20 cm
For Use With (Application)Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Includesn.a.
Measurement Time<3 sec.
ModelNanoDrop 2000c
No. of Samples1
Operating Temperature37°; ±0.5°C
Pathlength10, 5, 2, 1 mm
Power Consumption12 V DC
Sample Volume (Metric)1000, 0.5 to 2.0 μL
Spectral Resolution1.8 nm (FWHM at Hg 253.7)
System RequirementsMicrosoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
Volume (Metric)0.5 to 2.0 μL
Warranty1 year parts and labor
Wattage5 W
Wavelength Accuracy±1 nm
Weight (Metric)2.0 kg
Width (Metric)14 cm
Detector Type2048-element Linear Silicon CCD Array
Light SourceXenon Flash
TypeSpectrophotometer
Voltage12 VDC
Wavelength Range190 to 840 nm
Unit SizeEach

Frequently asked questions (FAQs)

How do I check the accuracy of my NanoDrop 2000/2000c Spectrophotometer?

Our NanoDrop CF-1 Calibration Fluid should be used with the Calibration Check module of the instrument software. CF-1 is prepared from the NIST potassium dichromate standard SRM935 in acidified reagent grade water (Certificate of Analysis available from NanoDrop products).

What additional analytes or biomolecules are routinely analyzed using the NanoDrop 2000/2000c Spectrophotometer?

Our NanoDrop 2000/2000c Spectrophotometers are routinely used to determine the concentrations of dsDNA, RNA, ssDNA, fluorescent dyes incorporated into microarray and proteomic probes, microbial cell cultures and virtually any analyte that exhibits absorbance in the 190-840 nm range.

Can I simply wipe the pedestal surface enough to prevent carryover on my NanoDrop 2000/2000c Spectrophotometer?

Yes, we recommend using a dry laboratory wipe as a very effective method for removing the sample. The highly polished quartz and stainless steel surfaces of the sample retention system are resistant to sample adherence.

What level of reproducibility should I expect from the NanoDrop 2000/2000c spectrophotometers?

The following is an example of the level of reproducibility that can be expected from the NanoDrop 2000/2000c Spectrophotometers:
BSA in A280 module:
- +/- 0.1 mg/mL for samples < 10 mg/mL
- +/- 2% for samples > 10 mg/mL

What sort of accuracy should I expect with the NanoDrop 2000/2000c spectrophotometers?

Typically, the accuracy of NanoDrop 2000/2000c Spectrophotometers falls within 2%.