NanoDrop™ 2000/2000c Spectrophotometer
Thermo Scientific™

NanoDrop™ 2000/2000c Spectrophotometer

이 모델은 더 이상 판매되지 않습니다. 최신 모델인 NanoDrop One Microvolume UV-Vis 분광광도계를 참조하십시오.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호유형포함
ND2000LAPTOPSpectrophotometerLaptop Computer
ND-2000Microvolume Spectrophotometern.a.
ND-2000CSpectrophotometern.a.
ND2000CLAPTOPUV-Vis Spec + Laptop ComputerLaptop Computer
카탈로그 번호 ND2000LAPTOP
제품 가격(KRW)
-
유형:
Spectrophotometer
포함:
Laptop Computer
이 모델은 더 이상 판매되지 않습니다. 최신 모델인 NanoDrop One Microvolume UV-Vis 분광광도계를 참조하십시오.

이 모델은 더 이상 판매되지 않습니다. 최신 모델인 NanoDrop One Microvolume UV-Vis 분광광도계를 참조하십시오.

 

US 특허 6,628,382 및 6,809,826.
사양
흡수 정확도3% (at 0.74 Abs at 350 nm)
흡수 범위0 to 300 A
정확도0.002
인증/적합성UL/CSA and CE
호환성Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
농도2 ng/μL to 15,000 ng/μL (dsDNA) ng/μL
연결USB
깊이(미터법)20 cm
검출 한계2 ng/μL (dsDNA) ng/μL
검출 범위2 to 15,000 ng/μL (dsDNA), 0.10 - 400mg/mL (BSA)
풋프린트14 x 20 cm
용도(애플리케이션)Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
포함Laptop Computer
인터페이스Laptop Computer
측정 시간<5 sec.
모델NanoDrop 2000 with laptop computer
샘플 수1
경로 길이0.05, 0.1, 0.2, and 1.0 mm
전력 소비12 V DC
샘플 부피(미터법)0.5 to 2.0 μL
스펙트럼 분해능1.8 nm (FWHM at Hg 253.7)
시스템 요구 사항Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
부피(미터법)0.5 to 2.0 μL
워런티1 year parts and labor
와트5 W
파장 정확도±1 nm
중량(미터법)2 kg
폭(미터법)14 cm
검출기 유형2048-element Linear Silicon CCD Array
광원Xenon Flash
제품라인Microvolume
스펙트럼 대역폭180 to 240 nm
유형Spectrophotometer
전압12 VDC
Wavelength Range190 to 840 nm
Unit Size-

자주 묻는 질문(FAQ)

How do I check the accuracy of my NanoDrop 2000/2000c Spectrophotometer?

Our NanoDrop CF-1 Calibration Fluid should be used with the Calibration Check module of the instrument software. CF-1 is prepared from the NIST potassium dichromate standard SRM935 in acidified reagent grade water (Certificate of Analysis available from NanoDrop products).

What additional analytes or biomolecules are routinely analyzed using the NanoDrop 2000/2000c Spectrophotometer?

Our NanoDrop 2000/2000c Spectrophotometers are routinely used to determine the concentrations of dsDNA, RNA, ssDNA, fluorescent dyes incorporated into microarray and proteomic probes, microbial cell cultures and virtually any analyte that exhibits absorbance in the 190-840 nm range.

Can I simply wipe the pedestal surface enough to prevent carryover on my NanoDrop 2000/2000c Spectrophotometer?

Yes, we recommend using a dry laboratory wipe as a very effective method for removing the sample. The highly polished quartz and stainless steel surfaces of the sample retention system are resistant to sample adherence.

What level of reproducibility should I expect from the NanoDrop 2000/2000c spectrophotometers?

The following is an example of the level of reproducibility that can be expected from the NanoDrop 2000/2000c Spectrophotometers:
BSA in A280 module:
- +/- 0.1 mg/mL for samples < 10 mg/mL
- +/- 2% for samples > 10 mg/mL

What sort of accuracy should I expect with the NanoDrop 2000/2000c spectrophotometers?

Typically, the accuracy of NanoDrop 2000/2000c Spectrophotometers falls within 2%.