NanoDrop Eight UV-Vis Spectrophotometer
NanoDrop Eight UV-Vis Spectrophotometer
NanoDrop Eight UV-Vis Spectrophotometer
NanoDrop Eight UV-Vis Spectrophotometer
NanoDrop Eight UV-Vis Spectrophotometer

NanoDrop Eight UV-Vis Spectrophotometer

Increase the chances of success in your analyses with efficient analysis of your sample quantity and quality using the Thermo Scientific™ NanoDrop™ Eight UV-Vis spectrophotometer. Ideal for pharma/biotech and academic researchers measuring larger number of nucleic acid and protein samples.
Have Questions?
Catalog NumberType
NDE-GLMicrovolume UV-Vis spectrophotometer
Catalog number NDE-GL
Price (USD)
16,503.50
Special offer
Online exclusive
Ends: 04-Dec-2026
25,390.00
Save 8,886.50
Each
Type:
Microvolume UV-Vis spectrophotometer
Price (USD)
16,503.50
Special offer
Online exclusive
Ends: 04-Dec-2026
25,390.00
Save 8,886.50
Each
Increase the chances of success in your analyses with efficient analysis of your sample quantity and quality using the Thermo Scientific™ NanoDrop™ Eight UV-Vis spectrophotometer. Ideal for pharma/biotech and academic researchers measuring larger number of nucleic acid and protein samples. NanoDrop Eight measures 8 samples at a time in less than 20 seconds. Software identifies sample contaminants and reports true sample concentration with the built-in Thermo Scientific™ Acclaro™ Sample Intelligence technology. Optional software allows you to more easily comply with 21 CFR Part 11 regulation.

Quantify your samples with confidence using the NanoDrop Eight multichannel spectrophotometer. This is the next generation NanoDrop 8000 with improved hardware and the best software features from the Thermo Scientifict™NanoDrop™ One/OneC spectrophotometer.


Acclaro Contaminant Identification ensures samples are high quality
Know true concentration of your nucleic acid or protein without the interfering effects of sample contaminants. Understanding your sample composition helps downstream experiments go right the first time.
• Contaminant identification – recognizes common contaminants in nucleic acid and protein samples that could inhibit downstream reactions or falsely elevate calculated concentrations
• Corrected calculations – applies chemometric algorithms to determine the true concentration of your analyte without the contaminant
• Differentiate DNA from RNA – algorithms can identify mammalian RNA contaminating a dsDNA sample or mammalian dsDNA contaminating an RNA sample


NanoDrop Eight enhancements
• Wider dynamic range – measure higher concentration samples without dilution – up to 10,000 ng/μL dsDNA or 145 mg/mL IgG
• Auto-Measure and Auto-Blank functions – multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF
• Ergonomic ambidextrous pipetting – Pedestal array is aligned perpendicular to the front of the instrument creating a natural pipetting motion for users; the pipette guide is magnetic and
movable from side-to-side allowing left- and righthanded users to quickly and comfortably dispense samples
• Automated exports for LIMS – set a directory for data to be automatically exported; have your LIMS software sweep the folder to pull in NanoDrop Eight instrument data for archiving


Regulatory compliance made easy
Optional Thermo Scientific™ SciVault™ Software helps your lab stay compliant with federal data
regulations every step of the way.
• Streamline management – Optional SciVault Software integrates into NanoDrop Eight Software user interface
• Comply with US FDA 21 CFR Part 11 – Control user account access, apply digital signatures, and review audit logs
• Network multiple instruments – Install SciVault Software on a central computer to control privileges and view audit logs across multiple NanoDrop Eight Instruments in different labs
• Validate your systems – IQ/OQ documents and services available to confirm your instrument is running within manufacturer’s specifications


Software updates to make your time more productive
• Ready to start – NanoDrop Eight Software comes hard-coded with the most frequently used measurement applications including dsDNA, RNA, Protein A280, and UV-Vis
• Always improving – user input keeps NanoDrop Software constantly moving forward with updates that deliver greater capabilities and efficiencies in the lab. Visit thermoscientific.com/nanodropsw to see and download the latest software applications. Register your instrument there as well so you won’t miss any enhancements.

Specifications
Absorbance Accuracy3% at 0.97 A, 302 nm, 23°C ± 2°C
Absorbance Range0.04 to 200 Abs (10 mm equivalent)
DescriptionNanoDrop Eight UV-Vis Spectrophotometer
Detection Limits2.0 to 10,000 ng/μL dsDNA; 0.03 - 145 mg/mL IgG
Dimensions (L x W x H)24 cm x 33 cm x 17 cm (9.4 in x 13 in x 6.7 in)
Measurement Range8 samples in ≤ 20 sec
Pathlength1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting)
Power Consumption3W at Idle; 15W at Working Conditions
Sample Volume (Metric)1 to 2 μL
Spectral Resolution≤ 1.8 nm (FWHM at Hg 254 nm)
Warranty1 Year
Wavelength Accuracy± 1 nm
Weight (English)8.2 lb.
Weight (Metric)3.7 kg
Detector Type2048-element CMOS Linear Image Sensor
Light SourceXenon Flash
Product LineNanoDrop Eight
TypeMicrovolume UV-Vis spectrophotometer
Voltage100/240 V
Wavelength Range190 to 850 nm
Unit SizeEach

Frequently asked questions (FAQs)

What are the computer requirements for NanoDrop Eight software?

PC requirements are covered in the NanoDrop Eight user guide, and the NanoDrop Eight software is compatible with Windows 10 (Professional and Enterprise) and Windows 11 (Professional and Enterprise).

How do I obtain service for my NanoDrop Eight?

For service in the United States and Canada, please call 877-724-7690 option 3 or email nanodrop@thermofisher.com.
For international service, please contact your local NanoDrop distributor.

Does NanoDrop Eight come with 21 CFR Part 11 compliance software?

Optional SciVault Software installs on a companion PC and gives labs the tools they need to comply with US FDA 21 CFR Part 11. This software allows users to manage user accounts, control user account access, apply digital signatures, and maintain audit logging.

How do I verify performance of my NanoDrop Eight?

Use the instrument’s performance verification diagnostic along with the PV-8 Performance Verification kit as described in the NanoDrop Eight user guide. Regular checks help ensure inter-channel consistency.

What are common sample-loading tips needed for NanoDrop Eight?

Our website provides a sample-loading technical note: ensure consistent pipetting, avoid bubbles, keep sample pedestals clean, and work quickly to reduce evaporation. Visit www.thermofisher.com/nanodrop