Do you offer staining trays for staining protein gels?
We offer the following staining trays for staining protein gels:
- StainEase Staining Trays (Cat. No. NI2400) for convenient and even staining of Mini and Midi gels. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Each tray easily fits up to 4 mini gels.
- Mini Gel Incubation Trays (Cat. No. 22843) for staining mini gels and western blot membranes.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
- StainEase Staining Trays (Cat. No. NI2400) for convenient and even staining of Mini and Midi gels. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Each tray easily fits up to 4 mini gels.
- Mini Gel Incubation Trays (Cat. No. 22843) for staining mini gels and western blot membranes.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.