NuPAGE™ MOPS SDS Running Buffer (Powder)
Invitrogen™

NuPAGE™ MOPS SDS Running Buffer (Powder)

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to resolve medium- to large-sized proteins by providing slower SDS-PAGE separations. Supplied as a powder for flexible preparation, room temperature storage, and a long 2-year shelf life.
Have Questions?
Catalog NumberQuantity
NP0002055 Packets
Catalog number NP000205
Price (ZAR)
689.40
Each
Quantity:
5 Packets
Price (ZAR)
689.40
Each

Invitrogen NuPAGE MOPS SDS Running Buffer (Powder) is formulated for use with Bis-Tris gels to improve the resolution and band sharpness of medium- to large-sized proteins. Compared to MES-based buffers, the MOPS buffer system supports reduced protein migration rates for enhanced separation performance. Supplied as a dry powder, this running buffer enables flexible preparation, room temperature storage, a two-year shelf life, and is designed to be a convenient and reliable choice for routine electrophoresis workflows.

Features and benefits include:

  • Improved resolution of medium- to large-sized proteins—slower protein migration with MOPS buffer enhances separation and band sharpness compared to faster MES-based buffers
  • Optimized for Bis-Tris gel workflows—designed for use with Bis-Tris gels including both NuPAGE Bis-Tris and Bolt Bis-Tris Plus gels
  • Powder format for flexible preparation—prepare fresh running buffer as needed to support consistent electrophoresis performance and simplify laboratory workflows
  • Room temperature storage and extended shelf life—powder format enables convenient storage with a two-year shelf life for inventory efficiency
  • Reliable performance for routine SDS-PAGE—supports consistent and reproducible results across common protein electrophoresis applications when used under recommended conditions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)SDS-PAGE
For Use With (Equipment)Electrophoresis Chambers
FormulationMOPS SDS Running Buffer
Quantity5 Packets
Shelf Life2 Years
Storage RequirementsRoom Temperature
Sufficient ForPreparing 5 L Running Buffer
ColorWhite
FormatPowder
Gel TypeBis-Tris
Product LineNuPAGE
Product TypeSDS Running Buffer
Unit SizeEach