Onstage Incubator for CellInsight CX5/NXT HCS Platforms
Thermo Scientific™
Onstage Incubator for CellInsight CX5/NXT HCS Platforms
The Invitrogen Onstage Incubator for CellInsight NXT/CX5 High Content Screening (HCS) Platforms is an environmental chamber designed specifically CellInsight NXTRead more
Have Questions?
Catalog NumberQuantity
NX5LIVE0021 incubator
Catalog number NX5LIVE002
Price (USD)
46,630.00
Each
Quantity:
1 incubator
Price (USD)
46,630.00
Each
The Invitrogen Onstage Incubator for CellInsight NXT/CX5 High Content Screening (HCS) Platforms is an environmental chamber designed specifically CellInsight NXT and CellInsight CX5 series platforms. As an optional module, it enables precise control of temperature, humidity, and three gases for time-lapse imaging of live cells under both physiological and non-physiological conditions. Environmental settings and image acquisition parameters are all seamlessly integrated into the HCS Studio software creating a powerful, walk-away platform for live cell quantitative imaging. The Onstage Incubator for CellInsight NXT/CX5 HCS Platforms can be upgraded at any time to existing compatible units.
Advantages of the Onstage Incubator for CellInsight NXT/CX5 HCS Platforms:
• Fully integrated environmental chamber for live cell time-lapse imaging
• Precisely maintain physiological or non-physiological conditions
• Simple scheduling to define frequency of imaging over a time course
• Powerful kinetic and motility measurements that increase information gathered from each cell over time
• Field upgradeable
• Compatible with fixed endpoint screening using the Thermo Scientific Orbitor RS Plate Mover
Requirements
The Onstage Incubator for CellInsight NXT/CX5 HCS Platforms requires the latest version of HCS Studio software and STore Image and Data Management Software.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 incubator
Shipping ConditionRoom Temperature
Product LineCellInsight
Unit SizeEach