Oregon Green™ 488 Cadaverine, 5-isomer
Invitrogen™
Oregon Green™ 488 Cadaverine, 5-isomer
Oregon Green™ 488 Cadaverine, 5-isomer
Have Questions?
Catalog NumberQuantity
O10465
also known as O-10465
5 mg
Catalog number O10465
also known as O-10465
Price (USD)
435.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
435.00
Each
The primary aliphatic amine of the green fluorescent Oregon Green™ cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This molecule can also be used as a water-soluble, fixable polar tracer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Label or DyeOregon Green™ 488
Product TypeCadaverine
Quantity5 mg
Reactive MoietyAmine, Cadaverine
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product LineOregon Green
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (2)
Citations & References
Abstract
Optical properties and application of a reactive and bioreducible thiol-containing tetramethylrhodamine dimer.
Authors:Christie RJ, Tadiello CJ, Chamberlain LM, Grainger DW,
Journal:Bioconjug Chem
PubMed ID:19249862
'Thiolated dimeric tetramethylrhodamine (TAMRA) was synthesized in a straightforward procedure utilizing commercially available 5(6)-succinimidyl TAMRA and cystamine hydrochloride. The thiol-containing TAMRA dimer displayed distinct spectral properties in reduced and oxidized forms; covalent dimer formation produced greater effects on the spectral properties than previously reported for noncovalent TAMRA dimers or dimers ... More
A facile synthetic method to prepare fluorescently labeled ROMP polymers.
Authors:Roberts KS, Sampson NS
Journal:Org Lett
PubMed ID:15355025
'[structure: see text] To probe the activities of sperm ADAM protein (fertilinbeta), we devised a general synthetic strategy to generate fluorescently labeled fertilinbeta oligopeptide polymers. Immunofluorescence studies with these polymers demonstrated that fertilinbeta polymers bind specifically to a protein receptor on the mouse egg plasma membrane.' ... More