Oregon Green™ 514 Carboxylic Acid, Succinimidyl Ester
Invitrogen™

Oregon Green™ 514 Carboxylic Acid, Succinimidyl Ester

Oregon Green™ 514 carboxylic acid, succinimidyl ester
Have Questions?
Catalog NumberQuantity
O61395 mg
Catalog number O6139
Price (USD)
443.00
Each
Quantity:
5 mg
Price (USD)
443.00
Each

The amine-reactive Oregon Green™ 514 succinimidyl ester can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼489/526 nm.

This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa, making its fluorescence essentially pH insensitive in the physiological pH range.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical ReactivityAmine
Emission526
Excitation489
Label or DyeOregon Green™ 514
Product TypeSuccinimidyl Ester
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product LineOregon Green
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (12)

Citations & References
Abstract
Real-time multiple particle tracking of gene nanocarriers in complex biological environments.
Authors:Lai SK, Hanes J,
Journal:Methods Mol Biol
PubMed ID:18470640
'Complex biological fluids, such as the vast and molecularly crowded cell cytoplasm and the highly viscoelastic mucus that protects many entry ways to the body, pose significant barriers to efficient gene delivery. Understanding the dynamics of gene carriers in such environments allows insight that leads to rational improvements in gene ... More
A mismatch-selective bifunctional rhodium-Oregon Green conjugate: a fluorescent probe for mismatched DNA.
Authors:Zeglis BM, Barton JK,
Journal:J Am Chem Soc
PubMed ID:16637630
A mismatch-selective bifunctional rhodium-Oregon Green conjugate: a fluorescent probe for mismatched DNA. Zeglis BM, Barton JK. Division of Chemistry and Chemical Engineering, California Institute of Technology, Pasadena, California 91125, USA. Publication Types: Research Support, N.I.H., Extramural Research Support, U.S. Gov't, Non-P.H.S. PMID: 16637630 [PubMed - indexed for MEDLINE] ... More
Synthesis and characterization of fluorescent cobalamin (CobalaFluor) derivatives for imaging.
Authors:Smeltzer CC, Cannon MJ, Pinson PR, Munger JD, West FG, Grissom CB
Journal:Org Lett
PubMed ID:11263885
Fluorescent derivatives of cobalamin have been prepared by linking fluorophores to cobalamin through a propylamide spacer. Fluorescein, naphthofluorescein, and Oregon Green derivatives have been prepared in good yield by reaction of the fluorophore NHS-ester with beta-(3-aminopropyl)cobalamin to form fluorescent cobalamin conjugates (CobalaFluors) that are potentially suitable for the in vitro ... More
Fiber-optic nitric oxide-selective biosensors and nanosensors.
Authors:Barker SL, Kopelman R, Meyer TE, Cusanovich MA
Journal:Anal Chem
PubMed ID:9511472
Fiber-optic biosensors that are selective for nitric oxide and do not respond to most potential interferents have been prepared with cytochromes c'. Both micro- and nanosensors have been prepared, and their response is fast (< 1 s), reversible, and linear up to 1 mM nitric oxide. The detection limit is ... More
Novel immuno-FRET assay method for Bacillus spores and Escherichia coli O157:H7.
Authors:Bruno JG, Ulvick SJ, Uzzell GL, Tabb JS, Valdes ER, Batt CA
Journal:Biochem Biophys Res Commun
PubMed ID:11573946
Novel immunofluorescence resonance energy transfer (immuno-FRET) assays for both Bacillus cereus spores and Escherichia coli O157:H7 are reported. Both assays involve the use of dual (QSY-7 and Oregon Green 514-antibody)-labeled spores or vegetative bacteria, such that Oregon Green 514-labeled antibodies are quenched by proximal QSY-7 molecules that are covalently bound ... More