Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NMDAR1 Antibodies

          Invitrogen

          NMDAR1 Polyclonal Antibody

          View all (56) NMDAR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NMDAR1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          NMDAR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NMDAR1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NMDAR1 Antibody (OPA1-04012) in WB

          Western blot of NMDAR1 in HEK 293 cells (10 µg). Showing specific immunolabeling of a ~120 kDa band corresponding to NR1 subunit of the NMDAR1 polyclonal antibody (Product # OPA1-04012) receptor containing the C1 splice variant insert in lane 3. HEK cells without NR1 expression (lane 1); NR1 subunit containing only the C2 Insert (lane 2); NR1 subunit containing the C1 and C2 Insert (lane 3); NR1 subunit containing the N1 and C2 insert (lane 4). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NMDAR1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          OPA1-04012

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:1,000-1:2,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:1,000-1:2,000
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit of rat GRIN1, C1 splice variant insert conjugated to KLH

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.5 mg/mL once reconstituted

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          5mM Ammonium bicarbonate, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325647

          Product Specific Information

          This antibody is predicted to react with bovine, canine, human and non-human primate based on 100% sequence homology.

          In Western blot, this antibody detects an ~100 kDa protein representing the NR1 C1 splice variant from rat brain homogenate. It shows no cross-reactivity with the NR1 N1, C2, or C2 splice variants.

          Reconstitute with 50 µL of PBS.

          Target Information

          NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Glun1r; glutamate [NMDA] receptor subunit zeta-1; glutamate receptor; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDZ1; NMZ1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; RP11-350O14.1; unnamed protein product

          View more View less

          Gene Aliases: GluN1; GluRdelta1; GluRzeta1; M100174; NMD-R1; Nmdar; NMDAR1; NR1; Rgsc174

          View more View less

          UniProt ID: (Rat) P35439, (Mouse) P35438

          View more View less

          Entrez Gene ID: (Rat) 24408, (Mouse) 14810

          View more View less

          Function(s)
          beta-amyloid binding ionotropic glutamate receptor activity NMDA glutamate receptor activity receptor binding ion channel activity cation channel activity calcium channel activity calcium ion binding protein binding calmodulin binding ligand-gated ion channel activity ligand-gated sodium channel activity glycine binding glutamate binding enzyme binding phosphatase binding transmitter-gated ion channel activity voltage-gated cation channel activity glutamate-gated calcium ion channel activity glutamate receptor binding signaling receptor activity macromolecular complex binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential ionotropic glutamate receptor binding
          Process(es)
          conditioned taste aversion response to hypoxia startle response suckling behavior response to amphetamine ion transport cation transport calcium ion transport cellular calcium ion homeostasis positive regulation of cytosolic calcium ion concentration chemical synaptic transmission learning or memory learning memory long-term memory associative learning adult locomotory behavior olfactory learning visual learning positive regulation of calcium ion transport into cytosol regulation of cell communication response to amine propylene metabolic process sensory perception of pain calcium-mediated signaling pons maturation hypothalamus development cerebral cortex development social behavior ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic sodium ion transmembrane transport regulation of membrane potential positive regulation of apoptotic process response to morphine regulation of neuron apoptotic process negative regulation of neuron apoptotic process regulation of respiratory gaseous exchange response to ethanol positive regulation of transcription from RNA polymerase II promoter regulation of synaptic plasticity regulation of neuronal synaptic plasticity regulation of long-term neuronal synaptic plasticity regulation of dendrite morphogenesis regulation of axonogenesis regulation of synapse organization neuromuscular process protein heterotetramerization positive regulation of mitochondrial calcium ion concentration response to calcium ion regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic calcium ion homeostasis regulation of postsynaptic membrane potential excitatory postsynaptic potential prepulse inhibition male mating behavior response to fungicide protein-containing complex assembly calcium ion transmembrane transport cellular response to manganese ion calcium ion transmembrane import into cytosol cation transmembrane transport regulation of presynaptic membrane potential chemical synaptic transmission, postsynaptic regulation of plasma membrane bounded cell projection organization positive regulation of Schwann cell migration positive regulation of dendritic spine maintenance response to glycoside positive regulation of reactive oxygen species biosynthetic process protein localization to postsynaptic membrane regulation of cation transmembrane transport cellular response to beta-amyloid response to glycine cellular response to glycine positive regulation of excitatory postsynaptic potential rhythmic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-hj2rq:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0