Xcalibur™ Software
Actual product may vary
Thermo Scientific™
Xcalibur™ Software
Thermo Scientific™ Xcalibur™ software lets you analyze samples from a single interface, including instrument setup, method development, data acquisition, and more.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
OPTON-30966Software4.3 Upgr fr 4.2/4.2 SP1
OPTON-30965Software4.3 + NIST Libraries
OPTON-30967Software4.3 Upgr fr 4.1 and older
Catalog number OPTON-30966
Price (USD)
-
Type:
Software
Includes:
4.3 Upgr fr 4.2/4.2 SP1
Control and process data from Thermo Scientific™ LC-MS systems and related instruments with easy-to-use Thermo Scientific™ Xcalibur™ software. Windows™-based Xcalibur software provides method setup, data acquisition, data processing, and data acquisition and processing integration to provide higher quality and more relevant data. Xcalibur software integrates seamlessly with application-specific Thermo Scientific software packages, and now includes both Thermo Scientific™ FreeStyle™ and Thermo Scientific™ Almanac™ web-based monitoring for cloud-enabled and cloud-independent functionality.
Efficient, simple data acquisition, interrogration and in-depth analysis
Xcalibur software provides efficient data acquisition, data processing, and results delivery, all via an easy-to-use interface.
  • Access your comprehensive toolset through the homepage, from included applications such as FreeStyle and Quan Browser to optional application-specific data processing tools to go seamlessly from method development to data acquisition, processing, and reporting
  • Ensure that you are always up-to-date with notifications when new version of software are available
  • Benefit from the web-based monitoring provided by cloud-based applications such as Almanac web-based monitoring to allow remote-monitoring of your systems and increase productivity
Specifications
For Use With (Equipment)Mass Spectrometers
Includes4.3 Upgr fr 4.2/4.2 SP1
Software4.3
TypeSoftware
Unit SizeEach