Owl™ Dual-Gel Vertical Electrophoresis Complete System, 20 x 20cm, 1150mL buffer volume - FAQs
Owl™ Dual-Gel Vertical Electrophoresis Complete System, 20 x 20cm, 1150mL buffer volume - FAQs
View additional product information for Owl™ Dual-Gel Vertical Electrophoresis Systems - FAQs (P10DS-3, P8DS-1, P8DS-2, P8DS-3, P10DS-1, P10DS-2)
1 product FAQs found
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.