Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Penta-His Tag Antibodies

          Invitrogen

          Penta-His Tag Monoclonal Antibody

          Advanced Verification
          5 Published Figures
          9 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Penta-His Tag Monoclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (5)
          • Advanced Verification (1)
          Penta-His Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Penta-His Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Penta-His Tag Antibody (P-21315) in WB

          Detection of an oligohistidine fusion protein with the Penta-His mouse monoclonal antibody. Twofold serial dilutions of an Escherichia coli lysate containing overexpressed oligomycin sensitivity-conferring protein (OSCP) fused with a hexahistidine domain were run an SDS-polyacrylamide gel and blotted onto a PVDF membrane. The blot was stained with the SYPRO Ruby protein blot stain (Product # S-11791) to detect the entire protein profile (top). After imaging, the blot was incubated with Penta-His mouse IgG1 monoclonal antibody (P... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Penta-His Tag Antibody in Western Blot (WB)
          Penta-His Tag Antibody in Western Blot (WB)
          Penta-His Tag Antibody in Western Blot (WB)
          Penta-His Tag Antibody in Western Blot (WB)
          Penta-His Tag Antibody in Western Blot (WB)
          Penta-His Tag Antibody in Immunoprecipitation (IP)
          Penta-His Tag Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          P-21315

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          View 7 publications 7 publications

          Immunohistochemistry (IHC)

          1:20-1:50
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:50
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Immunogen

          Histidine Tag (5xHis)

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS with PEG, sucrose

          Contains

          0.08% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2539819

          Product Specific Information

          Sufficient material is supplied to stain 50-100 minigel blots (8 cm × 10 cm). For immunodetection of oligohistidine fusion proteins on blotting membranes, use the Penta-His antibody at a final concentration of between 0.1 µg/mL and 0.2 µg/mL (1:1,000 to 1:2,000 dilution of the stock solution). Standard buffers and blocking agents, such as 3% BSA (bovine serum albumin) may be used; however, milk powder should not be used, as it reduces sensitivity. Denaturation of the proteins is recommended to ensure exposure of the oligohistidine domain. Detect the Penta-His antibody with an antimouse IgG secondary antibody conjugated to either alkaline phosphatase or horseradish peroxidase. The enzyme activity can then be visualized using a chromogenic, fluorogenic or chemiluminescent substrate.

          Upon receipt, the lyophilized antibody should be stored at 4°C. When properly stored, this product is stable for at least one year. To prepare a stock solution, reconstitute the antibody in 0.5 mL of deionized water (dH2O) to a final concentration of 0.2 mg/mL. Store the solution for up to 3 months at 4°C. For storage up to 6 months, divide the solution into single-use aliquots and freeze at -20°C. AVOID REPEATED FREEZING AND THAWING.

          Immunohistochemical methods can be used to investigate the location of recombinant proteins expressed in particular cells within tissues or organs, as well as for determination of the subcellular localization of proteins after targeting to particular cell compartments. The Penta-His antibody has been used to detect a hexahistidine aggrecan fusion protein in the perinuclear Golgi and the endoplasmic reticulum of transiently transfected CHO cells. The technique can also be used to localize cellular proteins that interact with hexahistidine fusion proteins. Preparation of the cells or tissue: Fix the tissue with 2% paraformaldehyde and permeabilize with 0.25% Triton™ X-100. This relatively mild fixation protocol best preserves epitopes and best maintains the position of the protein within the cellular structure. Immunodetection for cells and tissues: Before applying the antibody to the tissue, block nonspecific binding sites using 5% bovine serum albumin (BSA). For immunodetection of the oligohistidine fusion protein, use the Penta-His antibody at a final concentration of 410 µg/mL (1:20 to 1:50 dilution of the stock solution). Detect the Penta-His antibody with an anti-mouse IgG secondary antibody conjugated to a fluorescent dye, alkaline phosphatase, or horseradish peroxidase. The fluorescent dye can be seen directly using a fluorescence-based microscope. The enzyme activity can be detected using a chromogenic or fluorogenic substrate.

          Target Information

          The oligohistidine domain is a Ni2+-binding peptide sequence comprising a string of four to six histidine residues. When the DNA sequence corresponding to the oligohistidine domain is fused in frame with a gene of interest, the resulting fusion protein can be easily purified using a nickel-chelating resin. The Penta-His monoclonal IgG1 antibody provides a sensitive method for specific detection of fusion proteins that have an oligohistidine domain comprising five or six consecutive histidine residues. The antibody does not recognize tetrahistidine domains or domains in which the histidine string is interrupted by another amino acid. The antibody binds to the oligohistidine domain regardless of the surrounding amino acid context and even when the group is partially hidden, although subtle differences in the amino acid context may change the sensitivity limit for a particular fusion protein. This antibody can be used to detect oligohistidine fusion proteins on western blots, dot blots or colony blots, or in tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 5Xhis; Histidine 5X; pentahistidine

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch