Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit - FAQs
Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit - FAQs
View additional product information for Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit - FAQs (P21875)
1 product FAQs found
I have stained my blot with Pro-Q Emerald 300/488 Glycoprotein Blot Stain. Can I also stain the blot with SYPRO Ruby Blot Stain for total protein detection?
Yes. We recommend staining with SYPRO Ruby Blot Stain after staining with Pro-Q Emerald 300 or 488 Glycoprotein Blot Stain.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.