May I store antibodies or other proteins labeled with the pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) in solutions containing sodium azide?
We do not recommend storing antibodies or other proteins labeled with the pHrodo Deep Red Antibody Labeling Kit (Cat. Nos. P35355, P35356) in solutions containing sodium azide, as pHrodo Deep Red is not stable in the presence of sodium azide. Consider using an alternative, non-azide-based anti-microbial.