pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry
Invitrogen™

pHrodo™ BioParticles™ Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry

Achieve faster and more accurate results in phagocytosis assays with pHrodo BioParticles conjugates and kit. pHrodo BioParticles conjugates fluoresce brightly in endosomes and lysosomes, do not require wash steps or quencher dye, can be multiplexed, and are used in flow cytometry, HCA, and HTS.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSpeciesColor
P35360ConjugateE. coliDeep Red
P35361ConjugateE. coliRed
A10025Phagocytosis KitE. coliRed
A10010ConjugateS. aureusRed
P35367ConjugateS. aureusGreen
P35365ConjugateS. cerevisiaeGreen
Catalog number P35360
Price (USD)
591.65
Online exclusive
636.00
Save 44.35
Each
Product Type:
Conjugate
Species:
E. coli
Color:
Deep Red
Price (USD)
591.65
Online exclusive
636.00
Save 44.35
Each

Achieve faster staining and more accurate results without the need for wash steps or quencher dye with pH-sensitive pHrodo BioParticles conjugates and phagocytosis kit for flow cytometry. pHrodo BioParticles conjugates for phagocytosis are non-fluorescent outside the cell at neutral pH but fluoresce brightly in acidic pH environments, such as those of endosomes and lysosomes. They can be used in imaging, HCA, and HTA applications. The pHrodo BioParticles phagocytosis kit offers rapid assessment of phagocytic activity in whole blood using flow cytometry.

The fluorescence of pHrodo dye dramatically increases as pH decreases from neutral to acidic, making it an ideal tool to study phagocytosis and its regulation by drugs and/or environmental factors. Outside the cell, the dye is non-fluorescent, which eliminates the need for wash steps and quencher dyes. Once internalized, the dye fluoresces brightly within the acidic environments of endosomes and lysosomes, enabling rapid assay development and certainty of results when investigating phagocytic pathways and their regulation by drugs and/or environmental factors. As such, pHrodo dye conjugates can be used in plate readers, fluorescence microscopy imaging, and flow cytometry applications.

The pHrodo Red S. aureus BioParticles Conjugate for Phagocytosis (Cat. No. A10010) offers fast and accurate assay results. pHrodo Red dye conjugates are non-fluorescent outside the cell but fluoresce bright red in phagosomes. The pHrodo Red E. coli BioParticles Conjugate for Phagocytosis (Cat. No. P35361) detects phagocytosis and endocytosis, offers reduced signal variability and improved timing in sensitive experiments, and can be used in multiplex applications with a wide variety of blue, green, and far-red dyes and reporters such as GFP, Fuo-4, calcein, NucBlue, CellEvent Caspase 3/7 green, Mitosox Green, and Mitotracker Deep Red, among many others. The optimal absorption and fluorescence emission maxima of the pHrodo Red BioParticles Conjugate is approximately 560/585 nm, respectively. However, the fluorophore is readily excited with the 488-nm argon-ion laser installed on most flow cytometers.

The pHrodo Red E. coli BioParticles Phagocytosis Kit (Cat. No. A10025) for flow cytometry is designed for rapid and convenient measurement of phagocytic activity in whole blood samples by flow cytometry. The kit includes all the reagents required for assessing particle ingestion and red blood cell lysis.

pHrodo Deep Red dye is a low-background pH sensor dye that shows no signal in neutral conditions and only fluoresces in acidic environments. pHrodo Deep Red dye enables better discrimination of internalized cargo from outside the cell because it has an approximate pKa of 5 and does not fluoresce until it enters the late endosome and lysosome. pHrodo Deep Red dye can be detected using a Cy5 fluorescent filter set and has been validated for use in a variety of applications, including flow cytometry, fluorescent microscopy, high content analysis (HCA), and high throughput screening (HTS). It can also be multiplexed with a wide variety of blue, green, and red dyes and reporters such as GFP and RFPs, Mitosox Green or Red, CellEvent Caspase 3/7 Green or Red, NucBlue, or TMR, among many others.

pHrodo Deep Red E. coli BioParticles Conjugate for Phagocytosis (Cat. No. P35360) is a no-wash, low background, fluorogenic reagent developed for the study of phagocytosis in a live cell system. This conjugate is non-fluorescent outside the cell but fluoresces dark red in phagosomes. The low pKa of the pHrodo Deep Red E. coli BioParticles Conjugate eliminates non-specific signal from non-internalized cargo and only turns on in late endosomes and lysosomes.

The pHrodo Green dye conjugates, like pHrodo Red and pHrodo Deep Red dye conjugates, offer faster and more accurate results than any other phagocytosis assay. pHrodo Green conjugates are non-fluorescent outside the cell at neutral pH but fluoresce bright green at acidic pH, such as that of phagosomes. Use the ready-made pHrodo Green S. aureus BioParticles Conjugate (Cat. No. P35367) and pHrodo Green Zymosan BioParticles Conjugate (Cat. No. P35365) for Phagocytosis in imaging, HCA, HTS, and flow applications.

These pHrodo conjugate products include enough reagent to perform 100 tests when using 100 μL in each well of a 96-well plate, and they can be multiplexed with a wide variety of blue, red, and far-red dye reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others.

pHrodo Red Zymosan BioParticles Conjugate (Cat. No. P35364) is also available, and pHrodo Green dye is available as a conjugate of E. coli BioParticles (Cat. No. P35366).

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
DescriptionpHrodo™ Deep Red E. coli BioParticles™ Conjugate for Phagocytosis
Detection MethodFluorescence
Dye TypepHrodo™ Deep Red
Excitation/Emission640/655
FormLyophilized Powder
Quantity5 x 2 mg
Reagent TypeFluorogenic
SpeciesE. coli
ColorDeep Red
For Use With (Application)Cell Analysis
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, High Content Analysis Instrument
Product TypeConjugate
Unit SizeEach
Contents & Storage
Store at –20°C, desiccate, and protect from light.

Frequently asked questions (FAQs)

What is the number of E. coli bioparticles per vial in the pHrodo BioParticles Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry (Cat. No. P35360)?

1 vial with 2 mg contains ~ 6 x 10∧8 particles.

Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

Can I store reconstituted pHrodo BioParticles Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry?

Yes. Once reconstituted, pHrodo BioParticles Conjugates for Phagocytosis and Phagocytosis Kit, for Flow Cytometry (Cat. Nos. P35367, P35361, P35360, P35366, P35364, P35365, A10010) can be stored at 2 - 8 degrees C for several weeks, as long as sodium azide is added to a final concentration of 2 mM. If no sodium azide is added, the cell suspension needs to be used right away or on the same day to avoid contamination. DO NOT FREEZE the resuspended pHrodo bioparticle conjugates.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the concentration of bacterial particles for pHrodo BioParticles Conjugates for Phagocytosis?

The bacterial particles per weight for pHrodo BioParticles Conjugates for Phagocytosis, is as follows:

pHrodo E. Coli BioParticles Conjugate for Phagocytosis (Cat. Nos. P35360, P35361, A10025) contains 3 x 108 particles/mg.

pHrodo S. aureus BioParticles Conjugate for Phagocytosis (Cat. Nos. A10010, P35367) contains 3 x 108 particles/mg.

pHrodo Zymosan BioParticles Conjugate for Phagocytosis (Cat. No. P35365) contains 2 x 107 particles/mg.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use pHrodo Deep Red E. Coli Bioparticles Conjugate for Phagocytosis (Cat. No. P35360) with cell types other than RAW or MMM (J774A.1) cells?

Yes. You can use cell types for pHrodo Deep Red E. Coli Bioparticles Conjugate for Phagocytosis (Cat. No. 35360) other than RAW or MMM (J774A.1). The final concentration of BioParticles per cell and incubation time would have to be optimized if using other cell types. You may follow the protocol for the RAW and MMM cells and determine if any changes are required for your cells of interest.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.