pHrodo™ Antibody Labeling Kits
Invitrogen™

pHrodo™ Antibody Labeling Kits

Perform antibody internalization, endocytosis, and phagocytosis experiments with confidence and ease using the updated pH-sensitive, brightly fluorescing dyes and ready-to-use spin columns in the pHrodo Red and Green Antibody Labeling Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityExcitation/EmissionLabel or Dye
P360143 labelings kit560/585pHrodo Red
P353553 x 100 μg640/655 nmpHrodo Deep Red
P353563 x 1 mg640/655 nmpHrodo Deep Red
P360203 x 100 μg560/585 nmpHrodo Red
P360213 x 1 mg560/585 nmpHrodo Red
P360223 x 100 μg505/525 nmpHrodo Green
P360233 x 1 mg505/525 nmpHrodo Green
P360153 labelings kit505/525 nmpHrodo Green
Catalog number P36014
Price (MXN)
-
Quantity:
3 labelings kit
Excitation/Emission:
560/585
Label or Dye:
pHrodo Red
Elevate biopharmaceutical research with pHrodo Antibody Labeling kits, designed to characterize antibody internalization, a critical step during antibody-drug conjugate research and development. Kits provide a simple, detailed protocol and advanced purification columns to easily label and purify antibodies and other large proteins. Amine-reactive pHrodo dyes covalently bind to antibodies and other proteins to produce bright, soluble conjugates whose fluorescence increases in acidic organelles including endosomes and lysosomes. Antibody internalization can be evaluated using flow cytometry, fluorescence microscopy, and high-content screening.

The pHrodo Red Antibody Labeling Kit and pHrodo Green Antibody Labeling Kit provide the reagents needed to easily label three purified protein or antibody samples with pHrodo Red or pHrodo Green dye. These kits contain amine-reactive, pH-sensitive pHrodo Red or Green STP ester dye that covalently binds to free lysines anywhere they occur in an antibody or protein.

The dyes included in this kit are improved versions of pHrodo Red and pHrodo Green dyes. They are optimized for the creation of bright and soluble bioconjugates that are used in the study of antibody internalization, endocytosis, phagocytosis, and for development of antibody-drug conjugates. These dyes are also more soluble than the original pHrodo Red and pHrodo Green dyes, making them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation. pHrodo Red and pHrodo Green dyes are non-fluorescent outside the cell, but fluoresce bright red or bright green as the pH of their surroundings becomes more acidic, such as those found inside of phagosomes and endosomes.

The pHrodo Red and Green Microscale Labeling Kits offer the following benefits:
• Convenience—Labeling typically takes one hour, with less than 30 minutes of hands-on time
• Versatility—Labelling can be performed on 12-150 kDa molecular weight proteins
• Customization—Create your own probes for studying antibody internalization, phagocytosis, and endocytosis

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
ColorRed
Excitation/Emission560/585
For Use With (Application)Cell Analysis
For Use With (Equipment)Flow Cytometer
Product LinepHrodo
Product TypeProtein Labeling Kit
Quantity3 labelings kit
Shipping ConditionRoom Temperature
Sufficient For3 Labels
Label or DyepHrodo Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C), desiccate, protect from light, and do not freeze.

Citations & References (4)

Citations & References
Abstract
LILRA2 selectively modulates LPS-mediated cytokine production and inhibits phagocytosis by monocytes.
Authors:Lu HK, Mitchell A, Endoh Y, Hampartzoumian T, Huynh O, Borges L, Geczy C, Bryant K, Tedla N
Journal:PLoS One
PubMed ID:22479404
'The activating immunoglobulin-like receptor, subfamily A, member 2 (LILRA2) is primarily expressed on the surface of cells of the innate immunity including monocytes, macrophages, neutrophils, basophils and eosinophils but not on lymphocytes and NK cells. LILRA2 cross-linking on monocytes induces pro-inflammatory cytokines while inhibiting dendritic cell differentiation and antigen presentation. ... More
A Common Genetic Variation in Langerin (CD207) Compromises Cellular Uptake of Staphylococcus aureus.
Authors:van Dalen R, Fuchsberger FF, Rademacher C, van Strijp JAG, van Sorge NM
Journal:J Innate Immun
PubMed ID:31141812
'Langerhans cells are key sentinel cells of the skin and mucosal lining. They sense microorganisms through their repertoire of pattern-recognition receptors to mount and direct appropriate immune responses. We recently demonstrated that human Langerhans cells interact with the Gram-positive pathogen Staphylococcus aureus through the Langerhans cell-specific receptor langerin (CD207). It ... More
High Affinity Promotes Internalization of Engineered Antibodies Targeting FGFR1.
Authors:Opalinski L, Szymczyk J, Szczepara M, Kucinska M, Krowarsch D, Zakrzewska M, Otlewski J
Journal:Int J Mol Sci
PubMed ID:29748524
Fibroblast growth factor receptor 1 (FGFR1) is a plasma membrane protein that transmits signals from the extracellular environment, regulating cell homeostasis and function. Dysregulation of FGFR1 leads to the development of human cancers and noncancerous diseases. Numerous tumors overproduce FGFR1, making this receptor a perspective target for cancer therapies. Antibody-drug ... More
Novel Silyl Ether-Based Acid-Cleavable Antibody-MMAE Conjugates with Appropriate Stability and Efficacy.
Authors:Wang Y, Fan S, Xiao D, Xie F, Li W, Zhong W, Zhou X,
Journal:Cancers (Basel)
PubMed ID:31288450
Antibody-drug conjugate (ADC) is a novel efficient drug delivery system that has been successfully used in clinical practice, and it has become a research hotspot in the anti-tumor drug field. Acid-cleavable linkers were first used in clinical ADCs, but their structural variety (e.g., hydrazone and carbonate) is still limited, and ... More