Phycobiliprotein bioconjugates, including antibody and other protein conjugates, can be made with the exceptionally bright phycobiliprotein, R-phycoerythrin..Read more
Have Questions?
Catalog Number
Quantity
P801
0.5 mL
Catalog number P801
Price (USD)
383.00
Each
Quantity:
0.5 mL
Request bulk or custom format
Price (USD)
383.00
Each
Phycobiliprotein bioconjugates, including antibody and other protein conjugates, can be made with the exceptionally bright phycobiliprotein, R-phycoerythrin..
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration4 mg/mL
Product TypeR-Phycoerythrin
Quantity0.5 mL
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Citations & References (79)
Citations & References
Abstract
CD5 antibodies increase intracellular ionized calcium concentration in T cells.
Authors:June CH,Rabinovitch PS,Ledbetter JA
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:3106489
The binding of a variety of monoclonal antibodies to the CD5 (T, gp67) pan T cell differentiation antigen has been shown to potentiate T cell proliferation. In this paper we show that CD5 monoclonal antibodies cause increased intracellular free calcium concentration ([Ca2+]i) in T cells. An increase in [Ca2+]i occurred ... More
Transcriptional activation of HLA-DR alpha by interferon gamma requires a trans-acting protein.
Authors:Blanar MA, Boettger EC, Flavell RA
Journal:Proc Natl Acad Sci U S A
PubMed ID:3133656
Stimulation of the human epithelial-like cell line, HeLa, with interferon gamma (IFN-gamma) induces steady-state levels of HLA-DR alpha mRNA. Using a sensitive RNase-mapping procedure, we detect induced HLA-DR alpha mRNA as early as 8 hr after IFN-gamma treatment; maximal accumulation occurs by 48 hr. Treatment with the protein synthesis inhibitor, ... More
TCR engagement of CD4+CD8+ thymocytes in vitro induces early aspects of positive selection, but not apoptosis.
Authors:Groves T, Parsons M, Miyamoto NG, Guidos CJ
Journal:J Immunol
PubMed ID:8977176
'Immature CD4/CD8 double-positive (DP) thymocytes expressing self MHC-restricted TCR are positively selected in response to TCR signals to survive and differentiate into functionally competent CD4 or CD8 single positive (SP) T cells. In contrast, DP precursors expressing autoreactive TCR are clonally deleted in response to TCR signals. We show here ... More
ATP-dependent association of nuclear proteins with isolated rat liver nuclei.
Authors:Imamoto-Sonobe N, Yoneda Y, Iwamoto R, Sugawa H, Uchida T
Journal:Proc Natl Acad Sci U S A
PubMed ID:3368451
'In vitro association of Xenopus nucleoplasmin and mammalian nonhistone chromosomal high mobility group 1 (HMG1) protein with nuclei isolated from rat liver was examined. Efficient association of nuclear proteins with isolated nuclei requires ATP, HCO3-, and Ca2+. Association occurred at 33 degrees C but not at 4 degrees C. ATP ... More
Antioxidant activity of metallothionein compared with reduced glutathione.
Authors:Miura T, Muraoka S, Ogiso T
Journal:Life Sci
PubMed ID:9155005
'The antioxidant activity of metallothionein (MT) was investigated. Metallothionein scavenged hydroxyl radicals (HO.) to protect DNA from the oxidative attack by microsomes. This scavenging activity of MT was approximately 50 times greater than reduced glutathione (GSH) on a molar basis. However, MT did not inhibit DNA damage induced by microsomes ... More