Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SynGAP Antibodies

          Invitrogen

          SynGAP Polyclonal Antibody

          Advanced Verification
          37 Published Figures
          40 References
          View all (11) SynGAP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SynGAP Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (37)
          • Advanced Verification (1)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SynGAP Antibody (PA1-046) in ICC/IF

          Immunofluorescence analysis of SynGAP was performed using 70% confluent log phase PC-12 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with SynGAP Rabbit Polyclonal Antibody (Product # PA1-046) at 2 µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product #... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SynGAP Antibody in Immunohistochemistry (IHC)
          SynGAP Antibody in Immunohistochemistry (IHC)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunocytochemistry (ICC/IF)
          SynGAP Antibody in Immunohistochemistry (IHC)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot, ChIP Assay (WB, ChIP)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody in Western Blot (WB)
          SynGAP Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SynGAP Polyclonal Antibody

          Product Details

          PA1-046

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 23 publications 23 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 9 publications 9 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GST fusion protein of a recombinant fragment, residues 947-1167, of rat SynGAP.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2287112

          Product Specific Information

          PA1-046 detects synaptic Ras-GTPase activating protein (SynGAP) from mouse and rat tissues.

          The PA1-046 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ~135 kDa protein representing SynGAP from mouse brain tissue homogenate. Immunocytochemical staining of SynGAP in rat hippocampal neurons with PA1-046 yields a pattern consistent with vesicular staining.

          PA1-046 immunogen is residues 947-1167 of rat SynGAP.

          Target Information

          Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZp761G1421; Neuronal R; neuronal RasGAP; neuronal synaptic RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; Ras/Rap GTPase-activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; synaptic ras GTPase-activating protein; C-terminal splice variant; synaptic ras GTPase-activating protein; N-terminal splice variant; synaptic ras-GAP; synaptic Ras-GAP 1

          View more View less

          Gene Aliases: Gm1963; MRD5; RASA5; SYNGAP

          View more View less

          UniProt ID: (Human) Q96PV0, (Mouse) F6SEU4

          View more View less

          Entrez Gene ID: (Human) 8831, (Mouse) 240057, (Rat) 192117

          View more View less

          Function(s)
          GTPase activator activity protein binding protein kinase binding
          Process(es)
          negative regulation of Ras protein signal transduction regulation of synaptic plasticity modulation of synaptic transmission maintenance of postsynaptic specialization structure regulation of intracellular signal transduction Ras protein signal transduction pattern specification process visual learning dendrite development receptor clustering regulation of MAPK cascade negative regulation of neuron apoptotic process regulation of long-term neuronal synaptic plasticity negative regulation of axonogenesis regulation of synapse structure or activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-hhljg:80/100.66.77.101:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0