Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha-1a Adrenergic Receptor Antibodies

          Invitrogen

          alpha-1a Adrenergic Receptor Polyclonal Antibody

          5 Published Figures
          27 References
          View all (15) alpha-1a Adrenergic Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite alpha-1a Adrenergic Receptor Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (5)
          alpha-1a Adrenergic Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          alpha-1a Adrenergic Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          alpha-1a Adrenergic Receptor Antibody (PA1-047) in ICC/IF

          Immunofluorescent analysis of alpha-1 Adrenergic Receptor (green) showing staining in the cytoplasm of NIH-3T3 cells (right) compared to a negative control without primary antibody (left). Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with an Alpha-1 Adrenergic Receptor polyclonal antib... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          alpha-1a Adrenergic Receptor Antibody in Immunocytochemistry (ICC/IF)
          alpha-1a Adrenergic Receptor Antibody in Immunocytochemistry (ICC/IF)
          alpha-1a Adrenergic Receptor Antibody in Immunocytochemistry (ICC/IF)
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry (IHC)
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry (IHC)
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry (IHC)
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          alpha-1a Adrenergic Receptor Antibody in Immunohistochemistry - Free Floating (IHC (Free))

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          alpha-1a Adrenergic Receptor Polyclonal Antibody

          Product Details

          PA1-047

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:400
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:1,000
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Fruit fly, Human, Mouse, Rat, Zebrafish

          Published species

          Cat, Human, Mouse, Non-human primate, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues K(339) F S R E K K A A K T(349) of the 3rd intracellular loop of human A1AR.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2273801

          Product Specific Information

          PA1-047 detects alpha-1 adrenergic receptor (A1AR) from human, mouse, rat, zebra fish (Danio rerio) and Drosophila samples.

          PA1-047 has been successfully used in Western blot, immunohistochemistry and immunofluorescence procedures. By Western blot, this antibody detects an ~60 kDa protein representing the A1AR from mouse kidney membrane preparations, mouse liver, HeLa cell lysates, zebra fish and drosophila samples. Immunofluorescence staining of A1AR in mouse kidney distal tubule cells with PA1-047 results in staining of the plasma membrane.

          The PA1-047 immunogen is a synthetic peptide corresponding to residues K(339) F S R E K K A A K T(349) of the 3rd intracellular loop of human A1AR. This sequence is 100% conserved in all A1AR subtypes examined. PA1-047 immunizing peptide (Cat. # PEP-216) is available for use in neutralization and control experiments.

          Target Information

          Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence. A1AR subtypes are found in numerous tissues and have been shown to be involved in the regulation of blood pressure due to changes in vascular tone and cardiac output. Effects on uterine contraction, hepatic glucose metabolism, heat shock protein 70 (HSP70), proto-oncogene expression, and mitogenesis have been linked to A1AR activation. Norepinephrine can increase nitric oxide synthase (NOS) levels in the hypothalamus by activating alpha-1 AR which may indirectly suppress the release of luteinizing hormone (LH), the androgens, estrogen, and progesterone.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ADA1; adrenergic alpha 1c receptor; adrenergic receptor alpha 1c subtype; adrenergic receptor, alpha 1a; adrenergic receptor, alpha 1c; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1Aa, receptor; adrenoceptor alpha 1A; alpha 1c-adrenoceptor subtype; alpha-1A adrenoceptor; alpha-1A adrenoreceptor; alpha-1C adrenergic receptor; Alpha1C-AR; G protein coupled receptor; GPCR; unnamed protein product

          View more View less

          Gene Aliases: adra1a; ADRA1C; ADRA1L1; ALPHA1AAR; fc18h12; wu:fc18h12

          View more View less

          UniProt ID: (Human) P35348, (Rat) P43140, (Mouse) P97718

          View more View less

          Entrez Gene ID: (Human) 148, (Rat) 29412, (Mouse) 11549, (Zebrafish) 798498

          View more View less

          Function(s)
          G-protein coupled receptor activity adrenergic receptor activity alpha1-adrenergic receptor activity protein binding protein heterodimerization activity
          Process(es)
          negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure positive regulation of heart rate by epinephrine-norepinephrine positive regulation of the force of heart contraction by epinephrine-norepinephrine positive regulation of systemic arterial blood pressure regulation of muscle contraction G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration cell-cell signaling adult heart development regulation of blood pressure response to xenobiotic stimulus response to hormone positive regulation of heart rate negative regulation of autophagy positive regulation of cardiac muscle hypertrophy regulation of vasoconstriction positive regulation of synaptic transmission, GABAergic negative regulation of Rho protein signal transduction organ growth response to cocaine positive regulation of MAPK cascade positive regulation of action potential positive regulation of vasoconstriction positive regulation of smooth muscle contraction regulation of cardiac muscle contraction positive regulation of cardiac muscle contraction cell growth involved in cardiac muscle cell development positive regulation of ERK1 and ERK2 cascade adrenergic receptor signaling pathway adenylate cyclase-activating adrenergic receptor signaling pathway phospholipase C-activating adrenergic receptor signaling pathway pilomotor reflex presynaptic modulation of chemical synaptic transmission positive regulation of phospholipase C/protein kinase C signal transduction neuron-glial cell signaling signal transduction apoptotic process smooth muscle contraction negative regulation of cell proliferation intracellular signal transduction regulation of systemic arterial blood pressure by carotid sinus baroreceptor feedback vasoconstriction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch