Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Leptin Antibodies

          Invitrogen

          Leptin Polyclonal Antibody

          Advanced Verification
          13 Published Figures
          14 References
          View all (34) Leptin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Leptin Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (13)
          • Advanced Verification (2)
          Leptin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Leptin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Leptin Antibody (PA1-051) in ICC/IF

          Immunofluorescence analysis of Leptin was performed using 70% confluent log phase 3T3-L1 cells differentiated with StemPro Adipogenesis Supplement (Product # A10070-01) for 5 days. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Leptin Rabbit Polyclonal Antibody (Product # PA1-051) at 2µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Leptin Antibody in Immunocytochemistry (ICC/IF)
          Leptin Antibody in Immunohistochemistry (IHC)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Immunohistochemistry (IHC)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot, ELISA (WB, ELISA)
          Leptin Antibody in Western Blot, ELISA (WB, ELISA)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Western Blot (WB)
          Leptin Antibody in Immunomicroscopy (IM)
          Leptin Antibody in Immunomicroscopy (IM)
          Leptin Antibody
          Leptin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Leptin Polyclonal Antibody

          Product Details

          PA1-051

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1:50 - 1:200
          -

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          -

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          1.5 µg/1x10^6 cells
          -

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Sheep, Rat

          Published species

          Avian, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues Q(25) K V Q D D T K T L I K T I V T R I N D(44) of mouse Leptin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325786

          Product Specific Information

          PA1-051 detects leptin from human, mouse, sheep and rat samples.

          PA1-051 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry experiments. By Western blot, this antibody detects an ~16 kDa protein representing leptin from 3T3-L1 cell extract. Immunohistochemical staining of leptin in sheep brain with PA1-051 results in staining of the ventromedial hypothalamus.

          The PA1-051 immunogen is a synthetic peptide corresponding to residues Q(25) K V Q D D T K T L I K T I V T R I N D(44) of mouse Leptin. This sequence is completely conserved between mouse and bovine.

          Target Information

          Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD295 antigen; H-Leptin; HuB219; LEP-R; leptin (murine obesity homolog); leptin (obesity homolog, mouse); ob; OB-R; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; unnamed protein product

          View more View less

          Gene Aliases: LEPD; OB; obese; OBS

          View more View less

          UniProt ID: (Human) P41159, (Mouse) P41160, (Rat) P50596

          View more View less

          Entrez Gene ID: (Human) 3952, (Sheep) 443534, (Mouse) 16846, (Rat) 25608

          View more View less

          Function(s)
          DNA binding receptor binding hormone activity protein binding peptide hormone receptor binding leptin receptor binding cytokine activity peptide hormone
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis ovulation from ovarian follicle response to hypoxia positive regulation of cytokine production placenta development regulation of endothelial cell proliferation response to dietary excess cardiac muscle hypertrophy energy reserve metabolic process glycerol biosynthetic process lipid metabolic process phagocytosis signal transduction JAK-STAT cascade female pregnancy response to nutrient circadian rhythm regulation of blood pressure positive regulation of cell proliferation adult feeding behavior fatty acid catabolic process negative regulation of autophagy negative regulation of lipid storage response to activity sexual reproduction T cell differentiation response to nutrient levels positive regulation of TOR signaling negative regulation of appetite prostaglandin secretion response to estradiol positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production regulation of natural killer cell activation regulation of natural killer cell proliferation response to insulin response to vitamin E leptin-mediated signaling pathway positive regulation of luteinizing hormone secretion positive regulation of peroxisome proliferator activated receptor signaling pathway intracellular signal transduction bone mineralization involved in bone maturation negative regulation of appetite by leptin-mediated signaling pathway positive regulation of T cell proliferation regulation of natural killer cell mediated cytotoxicity positive regulation of protein import into nucleus negative regulation of apoptotic process positive regulation of ion transport positive regulation of MAPK cascade cellular response to leptin stimulus response to ethanol regulation of angiogenesis negative regulation of vasoconstriction negative regulation of glucose import positive regulation of JAK-STAT cascade positive regulation of insulin receptor signaling pathway regulation of bone remodeling positive regulation of follicle-stimulating hormone secretion positive regulation of developmental growth intestinal absorption leukocyte tethering or rolling regulation of nitric-oxide synthase activity negative regulation of transport regulation of cell cycle positive regulation of protein kinase B signaling regulation of lipoprotein lipid oxidation negative regulation of cartilage development negative regulation of glucagon secretion cellular response to L-ascorbic acid cellular response to retinoic acid regulation of brown fat cell differentiation energy homeostasis receptor signaling pathway via STAT bone growth positive regulation of cold-induced thermogenesis regulation of cytokine production involved in inflammatory response positive regulation of p38MAPK cascade positive regulation of fat cell apoptotic process activation of protein kinase C activity positive regulation of reactive oxygen species metabolic process negative regulation of glutamine transport positive regulation of hepatic stellate cell activation glucose metabolic process regulation of gluconeogenesis fatty acid beta-oxidation cholesterol metabolic process bile acid metabolic process determination of adult lifespan central nervous system neuron development insulin secretion regulation of intestinal cholesterol absorption positive regulation of protein modification process cellular response to insulin stimulus aorta development hormone metabolic process glucose homeostasis eating behavior regulation of fat cell differentiation regulation of hormone secretion regulation of lipid biosynthetic process regulation of catalytic activity regulation of insulin secretion regulation of steroid biosynthetic process elastin metabolic process adipose tissue development regulation of protein localization to nucleus negative regulation of insulin receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch