Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Coronin 1A Antibodies

          Invitrogen

          Coronin 1A Polyclonal Antibody

          Advanced Verification
          1 Published Figure
          1 Reference
          View all (19) Coronin 1A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Coronin 1A Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (1)
          • Advanced Verification (1)
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Coronin 1A Antibody (PA1-10022) in ICC/IF

          Immunofluorescent analysis of cortical neuron-glial cell culture from E20 rat stained with rabbit pAb to coronin 1a (Product # PA1-10022, diluted 1:1,000) in red, and costained with mouse mAb to GFAP (diluted 1:1,000) in green. The blue is Hoechst staining of nuclear DNA. The coronin 1a antibody labels protein expressed in the cytoplasm of microglia cells, while GFAP antibody stains intermediate filaments i... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Coronin 1A Antibody in Western Blot (WB)
          Coronin 1A Antibody in Western Blot (WB)
          Coronin 1A Antibody in Immunocytochemistry (ICC/IF)
          Coronin 1A Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Coronin 1A Polyclonal Antibody

          Product Details

          PA1-10022

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:4,000-1:10,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Pig, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          C-terminal peptide of human coronin 1a coupled to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          5mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2082069

          Product Specific Information

          PA1-10022 was made against the C-terminal peptide of human coronin 1a chemically coupled to keyhole limpet hemocyanin carrier protein. PA1-10022 is also known to work on many mammalian species both on western blots and on cells grown in tissue culture.

          Target Information

          This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Alternative splicing results in multiple transcript variants. A related pseudogene has been defined on chromosome 16.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: actin-binding; Clabp coronin; clipin-A; coronin 1; coronin, actin binding protein 1A; coronin, actin binding protein, 1A; coronin-1; coronin-like actin binding protein; coronin-like protein A; coronin-like protein p57; FLJ41407; MGC117380; TACO; tryptophan aspartate-containing coat protein; unnamed protein product

          View more View less

          Gene Aliases: CLABP; CLIPINA; HCORO1; IMD8; Lmb3; p57; TACO

          View more View less

          UniProt ID: (Human) P31146, (Bovine) Q92176, (Rat) Q91ZN1, (Mouse) O89053

          View more View less

          Entrez Gene ID: (Human) 11151, (Bovine) 282196, (Pig) 100621483, (Rat) 155151, (Mouse) 12721

          View more View less

          Function(s)
          RNA binding actin binding actin monomer binding protein binding cytoskeletal protein binding myosin heavy chain binding identical protein binding protein homodimerization activity phosphatidylinositol 3-kinase binding actin filament binding protein C-terminus binding non-motor actin binding protein
          Process(es)
          immunological synapse formation phagolysosome assembly phagocytosis actin filament organization cell migration actin cytoskeleton organization cell-substrate adhesion regulation of actin cytoskeleton organization natural killer cell degranulation innate immune response positive chemotaxis negative regulation of actin nucleation negative regulation of cellular component organization calcium ion transport regulation of cell shape positive regulation of cell migration leukocyte chemotaxis negative regulation of vesicle fusion uropod organization nerve growth factor signaling pathway positive regulation of T cell proliferation T cell homeostasis negative regulation of neuron apoptotic process homeostasis of number of cells within a tissue regulation of release of sequestered calcium ion into cytosol early endosome to recycling endosome transport cellular response to interleukin-4 regulation of actin polymerization or depolymerization biological_process T cell differentiation regulation of actin filament polymerization endocytic recycling response to cytokine negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of T cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-qh5rm:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0