Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Versican V0, V1 Neo Antibodies

          Invitrogen

          Versican V0, V1 Neo Polyclonal Antibody

          Advanced Verification
          33 Published Figures
          52 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Versican V0, V1 Neo Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (33)
          • Advanced Verification (2)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 36

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Versican V0, V1 Neo Antibody (PA1-1748A) in WB

          Western blot of versican in human aorta samples using Product # PA1-1748A. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC (PFA), ICC/IF)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC (PFA), ICC/IF)
          Versican V0, V1 Neo Antibody in Immunocytochemistry (ICC/IF)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC (PFA), ICC/IF)
          Versican V0, V1 Neo Antibody in Immunocytochemistry (ICC/IF)
          Versican V0, V1 Neo Antibody in Immunocytochemistry (ICC/IF)
          Versican V0, V1 Neo Antibody in Immunocytochemistry (ICC/IF)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Versican V0, V1 Neo Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Versican V0, V1 Neo Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Immunohistochemistry (IHC)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in Western Blot (WB)
          Versican V0, V1 Neo Antibody in ELISA (ELISA)
          Versican V0, V1 Neo Antibody
          Versican V0, V1 Neo Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Versican V0, V1 Neo Polyclonal Antibody

          Product Details

          PA1-1748A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 17 publications 17 publications

          Immunohistochemistry (IHC)

          -
          View 21 publications 21 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 4 publications 4 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          ELISA (ELISA)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C G G D(436) P E A A E(441) of human Versican V0/V1 Neo.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2304324

          Product Specific Information

          PA1-1748A detects versican from human samples.

          PA1-1748A has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~70 kDa protein representing versican from mouse cumulus oocyte complexes.

          PA1-1748A immunogen is a synthetic peptide corresponding to residues C G G D(436) P E A A E(441) of human Versican V0/V1 Neo. This sequence is completely conserved between mouse and rat. This immunizing peptide (PEP-235) can be used for neutralization and control experiments. This polyclonal antibody has been referred to as: JSCDPE or DPEAAE.

          Target Information

          Versican is a member of the family of large aggregating proteoglycans (also including aggrecan, brevican, and neurocan). The protein shows wide tissue distribution which includes fibrous, articular, and elastic cartilages, as well as nervous, epidermal, arterial, and loose connective tissues. Versican V1 has been shown to enhance cell proliferation, and also induce cell transformation and protect cells from apoptosis. This was demonstrated through its ability to downregulate the expression of proapoptotic Bad. Versican's existence as a extracellular matrix protein in human airways has also lead to the discovery of an inflammatory role in asthmatics.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: chondroitin sulfate proteoglycan core protein; chondroitin sulfate proteoglycan core protein 2; glial hyaluronate-binding protein; hyaluronan-binding chondroitin sulfate proteoglycan; large fibroblast proteoglycan; PGM; unnamed protein product; V1 Neo; versican proteoglycan; Versican V0; WGN 1

          View more View less

          Gene Aliases: CSPG2; ERVR; GHAP; PG-M; WGN; WGN1

          View more View less

          UniProt ID: (Human) P13611

          View more View less

          Entrez Gene ID: (Human) 1462

          View more View less

          Function(s)
          calcium ion binding protein binding glycosaminoglycan binding hyaluronic acid binding protein tyrosine phosphatase activator activity extracellular matrix structural constituent conferring compression resistance
          Process(es)
          skeletal system development osteoblast differentiation cell adhesion central nervous system development cell recognition glial cell migration system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch