Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UCP1 Antibodies

          Invitrogen

          UCP1 Polyclonal Antibody

          Advanced Verification
          53 Published Figures
          63 References
          View all (20) UCP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite UCP1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (53)
          • Advanced Verification (2)
          UCP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          UCP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 56

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UCP1 Antibody (PA1-24894) in WB

          Western blot was performed using Anti-UCP1 Polyclonal Antibody (Product # PA1-24894) and a 33 kDa band corresponding to UCP1 was observed only in Mouse Brown Adipose and not Mouse White Adipose. Tissue extracts (30 µg lysate) of Mouse Brown Adipose (Lane 1) and Mouse White Adipose (Lane 2) were electrophoresed using NuPAGE™ 4-12 % Bis-Tris gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001) by iBlot® 2 Dry Blotting System (Product # IB21001). The blot was probed with the pri... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (Paraffin), Immunocytochemistry (IHC (P), ICC/IF)
          UCP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Immunohistochemistry (IHC)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in Western Blot (WB)
          UCP1 Antibody in ChIP Assay (ChIP)
          UCP1 Antibody
          UCP1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          UCP1 Polyclonal Antibody

          Product Details

          PA1-24894

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 36 publications 36 publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:500
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Cat, Ferret, Human, Mouse, Rat

          Published species

          Human, Monkey, Mouse, Non-human primate, Rat, Rhesus monkey

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to human UCP1 sequence (amino acids 145-159) with N-terminal cysteine added, conjugated to KLH.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.2 mg/mL

          Amount

          120 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          15mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2241459

          Product Specific Information

          PA1-24894 detects UCP1 from human, mouse, and rat samples, and does not cross-react with UCP2 or UCP3. PA1-24894 is expected to cross react with bovine (93% conserved), canine (93% conserved), Macaque monkey (100% conserved), rabbit (93% conserved), and ovine (93% conserved) due to sequence homology.

          By Western Blot this antibody detects a band of approximately 32 kDa.

          Positive Control: Brown adipose tissue.

          Target Information

          Mitochondrial uncoupling proteins are members of the family of mitochondrial anion carrier proteins. UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: fat uncoupling protein; HGNC:12517; mitochondrial brown fat uncoupling protein 1-like; proton channel in the inner mitochondrial membrane; solute carrier family 25 member 7; thermogenin; UCP 1; uncoupling protein; uncoupling protein 1 (mitochondrial, proton carrier); uncoupling protein 1, mitochondrial; uncoupling protein, mitochondrial; unnamed protein product

          View more View less

          Gene Aliases: SLC25A7; UCP; Ucpa; Uncp

          View more View less

          UniProt ID: (Human) P25874, (Rat) P04633, (Mouse) P12242

          View more View less

          Entrez Gene ID: (Human) 7350, (Rat) 24860, (Mouse) 22227

          View more View less

          Function(s)
          GTP binding hydrogen ion transmembrane transporter activity oxidative phosphorylation uncoupler activity GDP binding transmembrane transporter activity purine ribonucleotide binding long-chain fatty acid binding anion binding cardiolipin binding protein binding
          Process(es)
          diet induced thermogenesis generation of precursor metabolites and energy regulation of transcription from RNA polymerase II promoter mitochondrial transport response to temperature stimulus response to cold response to nutrient levels cellular response to hormone stimulus cellular response to reactive oxygen species brown fat cell differentiation transmembrane transport cellular response to cold cellular response to fatty acid positive regulation of cold-induced thermogenesis hydrogen ion transmembrane transport regulation of reactive oxygen species biosynthetic process cellular response to dehydroepiandrosterone mitochondrial transmembrane transport adaptive thermogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch