Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRPV1 Antibodies

          Invitrogen

          TRPV1 Polyclonal Antibody

          3 References
          View all (48) TRPV1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TRPV1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          TRPV1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          TRPV1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRPV1 Antibody (PA1-29770) in IHC (F)

          Immunohistochemistry (Frozen) analysis of rat dorsal root ganglion using (Product # PA1-29770) TRPV1 Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRPV1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          TRPV1 Antibody in Immunohistochemistry (IHC)
          TRPV1 Antibody in Immunohistochemistry (IHC)
          TRPV1 Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TRPV1 Polyclonal Antibody

          Product Details

          PA1-29770

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Guinea pig / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          YTGSLKPEDAEVFKDSMVPGEK
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. not determined

          Storage buffer

          whole serum

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1958678

          Product Specific Information

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5' UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: capsaicin receptor; cation channel; deltaN-TRPV1; DKFZp434K0220; ion channel; ion channel subunit; osm-9-like TRP channel 1; OTRPC1; transient receptor potential cation channel V1; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; truncated capsaicin receptor protein; unnamed protein product; Vanilloid receptor; vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; vanilloid type 1 receptor; VR1; VR1 receptor

          View more View less

          Gene Aliases: OTRPC1; TRPV1_SON; TRPV1alpha; TRPV1beta; VR-1; VR.5'sv; VR1; Vr1l1

          View more View less

          UniProt ID: (Human) Q8NER1, (Rat) O35433, (Mouse) Q704Y3

          View more View less

          Entrez Gene ID: (Human) 7442, (Rat) 83810, (Mouse) 193034

          View more View less

          Function(s)
          transmembrane signaling receptor activity ion channel activity extracellular ligand-gated ion channel activity excitatory extracellular ligand-gated ion channel activity voltage-gated calcium channel activity calcium channel activity protein binding calmodulin binding ATP binding calcium-release channel activity phosphatidylinositol binding phosphoprotein binding temperature-gated ion channel activity cation channel activity cation transmembrane transporter activity ligand-gated ion channel activity identical protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ion transport cell surface receptor signaling pathway chemosensory behavior calcium-mediated signaling ion transmembrane transport cellular response to heat thermoception detection of temperature stimulus involved in sensory perception of pain protein homotetramerization transmembrane transport excitatory postsynaptic potential calcium ion transmembrane transport cellular response to alkaloid cellular response to ATP cellular response to acidic pH calcium ion import across plasma membrane response to capsazepine calcium ion import into cytosol temperature homeostasis fever generation microglial cell activation diet induced thermogenesis peptide secretion vascular process in circulatory system negative regulation of systemic arterial blood pressure lipid metabolic process calcium ion transport positive regulation of cytosolic calcium ion concentration blood circulation response to pH response to heat response to acidic pH negative regulation of heart rate negative regulation of mitochondrial membrane potential negative regulation of neuron projection development glutamate secretion urinary bladder smooth muscle contraction sensory perception of pain positive regulation of urine volume positive regulation of renal sodium excretion positive regulation of apoptotic process response to peptide hormone positive regulation of nitric oxide biosynthetic process positive regulation of vasoconstriction response to pain behavioral response to pain negative regulation of axon regeneration regulation of body fluid levels sensory perception of taste sensory perception of mechanical stimulus detection of temperature stimulus involved in thermoception detection of chemical stimulus involved in sensory perception of pain smooth muscle contraction involved in micturition cellular response to cytokine stimulus cellular response to tumor necrosis factor cellular response to growth factor stimulus cellular response to temperature stimulus negative regulation of establishment of blood-brain barrier positive regulation of sensory perception of pain positive regulation of membrane depolarization negative regulation of iodide transmembrane transport cellular response to nerve growth factor stimulus positive regulation of excitatory postsynaptic potential positive regulation of cardiac muscle cell differentiation system process cation transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch