Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Estrogen Receptor beta Antibodies

          Invitrogen

          Estrogen Receptor beta Polyclonal Antibody

          Advanced Verification
          37 Published Figures
          104 References
          View all (59) Estrogen Receptor beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Estrogen Receptor beta Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (37)
          • Advanced Verification (1)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 39

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Estrogen Receptor beta Antibody (PA1-311) in WB

          Western blot analysis was performed on nuclear enriched extracts (30 µg lysate) of LNCap (Lane 1), PC-3 (Lane 2), MCF-7 (Lane 3), MDA-MB-231 (Lane 4), T-47D (Lane 5) and HeLa (Lane 6). The blots were probed with Anti-Rabbit Polyclonal Antibody (Product # PA1-311, 1 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A270... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Estrogen Receptor beta Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Immunocytochemistry (ICC/IF)
          Estrogen Receptor beta Antibody in Immunocytochemistry (ICC/IF)
          Estrogen Receptor beta Antibody in Immunocytochemistry (ICC/IF)
          Estrogen Receptor beta Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Immunohistochemistry (IHC)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in Western Blot (WB)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody in ChIP Assay (ChIP)
          Estrogen Receptor beta Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Estrogen Receptor beta Polyclonal Antibody

          Product Details

          PA1-311

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 37 publications 37 publications

          Immunohistochemistry (IHC)

          -
          View 41 publications 41 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1 µg/mL
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1 µg/mL
          View 13 publications 13 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 6 publications 6 publications

          Gel Shift (EMSA)

          1 µg/mL
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Sheep, Rat

          Published species

          Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Sheep

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues A(55) E P Q K S P W C E A R S L E H(70) of rat ER beta.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325597

          Product Specific Information

          PA1-311 detects estrogen receptor (ER) beta from human, non-human primate, mouse, ovine and rat tissues.

          PA1-311 has been successfully used in Western blot, immunocytochemistry and gel shift procedures. By Western blot, this antibody detects an ~55-62 kDa protein representing ER beta from rat brain homogenate. PA1-311 efficiently supershifts rat ER beta/ER response element (vitellogenin xenopus A2 gene).

          The PA1-311 immunogen is a synthetic peptide corresponding to residues A(55) E P Q K S P W C E A R S L E H(70) of rat ER beta.

          Recombinant ER beta expressed in baculovirus system (Cat. # RP-311 and RP-312) can be purchased for control experiments in Western blot and gel shift assays.

          PA1-311 can be used with blocking peptide PEP-011.

          Target Information

          Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ER; ER beta; ER-beta; ERbeta 4; estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta 2; estrogen receptor beta 4; nuclear receptor; nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; splice variant; steroid hormone receptor; steroid receptor; unnamed protein product

          View more View less

          Gene Aliases: ER-BETA; ER[b]; ERB; Erb2; ERBETA; ESR-BETA; ESRB; ESTRB; NR3A2; ODG8

          View more View less

          UniProt ID: (Human) Q92731, (Sheep) Q9TU15, (Mouse) O08537, (Rat) Q62986

          View more View less

          Entrez Gene ID: (Human) 2100, (Sheep) 443066, (Mouse) 13983, (Rat) 25149

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding steroid binding protein binding zinc ion binding enzyme binding estrogen receptor activity estrogen response element binding sequence-specific DNA binding receptor antagonist activity transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding hormone binding peroxisome proliferator activated receptor binding heterocyclic compound binding estradiol binding steroid hormone binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter signal transduction cell-cell signaling negative regulation of cell growth intracellular estrogen receptor signaling pathway positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cellular response to estradiol stimulus ovarian follicle development behavioral fear response neuron migration epithelial cell development brain development learning or memory negative regulation of cell proliferation hormone-mediated apoptotic signaling pathway regulation of signal transduction intracellular steroid hormone receptor signaling pathway intracellular receptor signaling pathway cellular response to hormone stimulus response to testosterone regulation of cell proliferation vasodilation positive regulation of apoptotic process negative regulation of neuron apoptotic process estrous cycle positive regulation of epidermal growth factor receptor signaling pathway negative regulation of behavior negative regulation of smooth muscle cell proliferation negative regulation of epithelial cell proliferation Sertoli cell proliferation uterus development vagina development prostate gland epithelium morphogenesis epithelial cell maturation involved in prostate gland development negative regulation of androgen receptor signaling pathway positive regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide cellular response to lipid negative regulation of feeding behavior negative regulation of reactive oxygen species metabolic process male gonad development female gonad development response to xenobiotic stimulus response to hormone response to activity response to insecticide cerebellum development amygdala development hypothalamus development prostate gland development response to nutrient levels response to estradiol response to genistein response to nicotine response to estrogen response to human chorionic gonadotropin response to ethanol regulation of epithelial cell proliferation Sertoli cell development cellular response to magnetism cellular response to xenobiotic stimulus response to dexamethasone response to salt response to bisphenol A
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline