Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CYP1A1 Antibodies

          Invitrogen

          CYP1A1 Polyclonal Antibody

          4 References
          View all (16) CYP1A1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CYP1A1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CYP1A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CYP1A1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CYP1A1 Antibody (PA1-340) in ICC/IF

          Immunofluorescence analysis of Cytochrome P450 1A1 was done on 70% confluent log phase HepG2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Cytochrome P450 1A1 Rabbit Polyclonal Antibody (Product # PA1-340) at 1:250 dilution in0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CYP1A1 Antibody in Immunocytochemistry (ICC/IF)
          CYP1A1 Antibody in Western Blot (WB)
          CYP1A1 Antibody in Western Blot (WB)
          CYP1A1 Polyclonal Antibody

          Product Details

          PA1-340

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:250
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Purified, His-tagged, full length human P450 1A1 fusion protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325602

          Product Specific Information

          PA1-340 detects cytochrome P450 (P450) 1A1 from human cells. This antibody detects, to a lesser extent, other P450 isoforms.



          PA1-340 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~54 kDa protein representing P450 1A1 from MCF-7 cell extract.



          The PA1-340 immunogen is purified, His-tagged, full length human P450 1A1 fusion protein.

          Target Information

          This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: aryl hydrocarbon hydroxylase; CYP; CYP1A1v; alternatively spliced; CYPIA1; cytochrome P-450 protein; cytochrome P-450-1; cytochrome P1-450, dioxin-inducible; Cytochrome P450 1A1; Cytochrome P450 form 6; cytochrome P450, family 1, subfamily A, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; Cytochrome P450-C; Cytochrome P450-P1; flavoprotein-linked monooxygenase; functional drug metabolizing enzyme; Hydroperoxy icosatetraenoate dehydratase; P450; P450 form 6; P450-P1; unnamed protein product; xenobiotic monooxygenase

          View more View less

          Gene Aliases: AHH; AHRR; CP11; CYP1; CYP1A1; CYPIA1; P1-450; P450-C; P450DX

          View more View less

          UniProt ID: (Human) P04798

          View more View less

          Entrez Gene ID: (Human) 1543

          View more View less

          Function(s)
          catalytic activity monooxygenase activity iron ion binding protein binding arachidonic acid monooxygenase activity steroid hydroxylase activity oxidoreductase activity oxidoreductase activity, acting on diphenols and related substances as donors oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen flavonoid 3'-monooxygenase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen lyase activity oxygen binding enzyme binding heme binding Hsp70 protein binding demethylase activity metal ion binding Hsp90 protein binding vitamin D 24-hydroxylase activity estrogen 16-alpha-hydroxylase activity estrogen 2-hydroxylase activity long-chain fatty acid omega-hydroxylase activity hydroperoxy icosatetraenoate dehydratase activity long-chain fatty acid omega-1 hydroxylase activity
          Process(es)
          response to hypoxia long-chain fatty acid metabolic process liver development lipid hydroxylation lipid metabolic process fatty acid metabolic process steroid biosynthetic process steroid catabolic process porphyrin-containing compound metabolic process xenobiotic metabolic process steroid metabolic process estrogen metabolic process amine metabolic process toxin metabolic process response to xenobiotic stimulus response to nematode response to herbicide response to toxic substance ethylene metabolic process coumarin metabolic process flavonoid metabolic process response to iron(III) ion insecticide metabolic process dibenzo-p-dioxin metabolic process phenol-containing compound metabolic process dibenzo-p-dioxin catabolic process epoxygenase P450 pathway response to food response to lipopolysaccharide response to vitamin A response to genistein response to immobilization stress xenobiotic catabolic process vitamin D metabolic process hormone metabolic process retinol metabolic process long-chain fatty acid biosynthetic process 9-cis-retinoic acid biosynthetic process camera-type eye development nitric oxide metabolic process response to arsenic-containing substance digestive tract development tissue remodeling hydrogen peroxide biosynthetic process response to hyperoxia maternal process involved in parturition hepatocyte differentiation cellular response to copper ion omega-hydroxylase P450 pathway olefinic compound metabolic process positive regulation of G1/S transition of mitotic cell cycle response to diphenyl ether response to Aroclor 1254 response to 2,3,7,8-tetrachlorodibenzodioxine response to 3-methylcholanthrene
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline