Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BACE1 Antibodies

          Invitrogen

          BACE1 Polyclonal Antibody

          Advanced Verification
          49 Published Figures
          44 References
          View all (33) BACE1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BACE1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (49)
          • Advanced Verification (1)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 55

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BACE1 Antibody (PA1-757) in ICC/IF

          Immunofluorescent analysis of BACE1 (green) showing staining in the Golgi apparatus of Neuro-2a cells. Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a BACE1 polyclonal antibody (Product # PA1-757) in 3% BSA-PBS at a dilution of 1:100 and incubated overnight at 4 °C in a humidified chamber. Cells were washed with PBST and incubated with a DyLight-conjugated secondary antibody in PBS at room temperature in the dark.... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Immunocytochemistry (ICC/IF)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Western Blot (WB)
          BACE1 Antibody in Immunoprecipitation (IP)
          BACE1 Antibody in Immunoprecipitation (IP)
          BACE1 Antibody in ELISA (ELISA)
          BACE1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          BACE1 Polyclonal Antibody

          Product Details

          PA1-757

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:2,000
          View 32 publications 32 publications

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 6 publications 6 publications

          ELISA (ELISA)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C(485) L R Q Q H D D F A D D I S L L K(501) of human beta-secretase.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325863

          Product Specific Information

          PA1-757 detects transfected beta-secretase 1.

          PA1-757 has been successfully used in ELISA, ICC/IF, Western blot and immunoprecipitation applications. The molecular weight of BACE1 detected may vary depending on the lysate tested. This may be due to glycosylation differences in various cell lines.

          The PA1-757 immunizing peptide (Cat. # PEP-093) is available for use in neutralization and control experiments.

          Target Information

          BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides (Abeta) found in extracellular amyloid plaques of Alzheimer's disease (AD). In some cases early onset familial AD can be attributed to a "Swedish" mutation in the amyloid precursor protein (APP), which dramatically enhances the cleavage of this protein by BACE1. This and other genetic and pathological evidence has led to therapeutic approaches that have focused on the inhibition of BACE1 and other APP-cleaving enzymes, such as gamma-secretase stuff. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer's disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: APP beta-secretase; asp 2; aspartyl protease 2; BACE; BACE 1; BACE-1B; BACE-1C; BACE-1D; BACE-I-432; BACEdelta7; Beta secretase; beta-site amyloid beta A4 precursor protein-cleaving enzyme; beta-site amyloid precursor protein cleaving enzyme 1; beta-site APP cleaving enzyme 1; beta-site APP processing enzyme; beta-site APP-cleaving enzyme; Memapsin; memapsin-2; Memapsin2; membrane type aspartyl protease; membrane-associated aspartic protease 2; transmembrane aspartic proteinase Asp2; unnamed protein product

          View more View less

          Gene Aliases: ASP2; BACE; HSPC104

          View more View less

          UniProt ID: (Human) P56817, (Mouse) P56818

          View more View less

          Entrez Gene ID: (Human) 23621, (Mouse) 23821

          View more View less

          Function(s)
          beta-amyloid binding endopeptidase activity aspartic-type endopeptidase activity protein binding peptidase activity beta-aspartyl-peptidase activity enzyme binding protein serine/threonine kinase binding
          Process(es)
          proteolysis membrane protein ectodomain proteolysis memory response to lead ion protein processing beta-amyloid formation swimming behavior amyloid precursor protein catabolic process regulation of neuron apoptotic process positive regulation of neuron apoptotic process regulation of synaptic plasticity beta-amyloid metabolic process modulation of synaptic transmission detection of mechanical stimulus involved in sensory perception of pain prepulse inhibition cellular response to copper ion cellular response to manganese ion presynaptic modulation of chemical synaptic transmission signaling receptor ligand precursor processing cellular response to beta-amyloid response to insulin-like growth factor stimulus amyloid fibril formation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch