Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NCoR1 Antibodies

          Invitrogen

          NCoR1 Polyclonal Antibody

          Advanced Verification
          15 Published Figures
          28 References
          View all (12) NCoR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NCoR1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (15)
          • Advanced Verification (6)
          NCoR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCoR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 25

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NCoR1 Antibody (PA1-844A) in ICC/IF

          Immunofluorescent analysis of NCoR (green) showing staining in the nucleus and cytoplasm of MCF-7 cells. Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a NCoR polyclonal antibody (Product # PA1-844A) in 3% BSA-PBS at a dilution of 1:200 and incubated overnight at 4 °C in a humidified chamber. Cells were washed with PBST and incubated with a DyLight-conjugated secondary antibody in PBS at room temperature in the dar... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NCoR1 Antibody in Immunocytochemistry (ICC/IF)
          NCoR1 Antibody in Immunocytochemistry (ICC/IF)
          NCoR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCoR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCoR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCoR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCoR1 Antibody in Immunocytochemistry (ICC/IF)
          NCoR1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in Western Blot (WB)
          NCoR1 Antibody in ChIP Assay (ChIP)
          NCoR1 Antibody in ChIP Assay (ChIP)
          NCoR1 Antibody
          NCoR1 Antibody
          NCoR1 Antibody
          NCoR1 Antibody
          NCoR1 Antibody
          NCoR1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NCoR1 Polyclonal Antibody

          Product Details

          PA1-844A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          -

          ChIP assay (ChIP)

          Assay-dependent
          View 9 publications 9 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C P(2427) A P L L S A Q Y E T L S D S D D(2443) of mouse N-CoR.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.7 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2149004

          Product Specific Information

          PA1-844A detects human and mouse nuclear receptor corepressor (N-CoR).

          PA1-844A has been successfully used in Western blot, ICC/IF, IHC (P) and immunoprecipitation (ChIP) procedures. By Western blot, this antibody detects a protein at ~270 kDa representing N-CoR from HeLa cell extracts and mouse RAW 264.7/primary macrophages. This antibody is not recommended for mouse C2C12 cells in immunofluorescent applications.

          The PA1-844A immunogen is a synthetic peptide corresponding to residues C P(2427) A P L L S A Q Y E T L S D S D D(2443) of mouse N-CoR. This immunizing peptide (Cat. # PEP-061) is available for use in neutralization and control experiments.

          Target Information

          This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 17 and 20.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: h KIAA1047; hCIT529I10; MGC104216; N-CoR1; nrcr; nuclear receptor corepressor; protein phosphatase 1, regulatory subunit 109; putative; retinoid X receptor-interacting protein 13; thyroid hormone- and retinoic acid receptor-associated corepressor 1; unnamed protein product

          View more View less

          Gene Aliases: 5730405M06Rik; A230020K14Rik; hN-CoR; mKIAA1047; N-CoR; N-CoR1; PPP1R109; RIP13; Rxrip13; TRAC1

          View more View less

          UniProt ID: (Human) O75376

          View more View less

          Entrez Gene ID: (Human) 9611, (Mouse) 20185

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding transcription corepressor activity protein binding ligand-dependent nuclear receptor binding histone deacetylase binding thyroid hormone receptor binding RNA polymerase II sequence-specific DNA binding transcription factor binding RNA polymerase II regulatory region sequence-specific DNA binding DNA binding chromatin binding protein domain specific binding estrogen receptor binding histone deacetylase regulator activity retinoic acid receptor binding peroxisome proliferator activated receptor binding sequence-specific DNA binding retinoid X receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development regulation of thyroid hormone mediated signaling pathway CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation regulation of transcription from RNA polymerase II promoter positive regulation of cell proliferation epidermis development gene expression negative regulation of gene expression thalamus development forebrain development circadian regulation of gene expression regulation of multicellular organism growth cholesterol homeostasis locomotor rhythm negative regulation of glycolytic process negative regulation of transcription, DNA-templated negative regulation of fatty acid metabolic process negative regulation of JNK cascade protein stabilization spindle assembly negative regulation of protein kinase B signaling definitive erythrocyte differentiation negative regulation of androgen receptor signaling pathway establishment of skin barrier negative regulation of intracellular signal transduction negative regulation of pri-miRNA transcription from RNA polymerase II promoter cellular response to Thyroglobulin triiodothyronine regulation of cellular ketone metabolic process negative regulation of transcription initiation from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch