Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PMCA2 ATPase Antibodies

          Invitrogen

          PMCA2 ATPase Polyclonal Antibody

          Advanced Verification
          39 Published Figures
          46 References
          View all (7) PMCA2 ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PMCA2 ATPase Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (39)
          • Advanced Verification (3)
          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 44

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PMCA2 ATPase Antibody (PA1-915) in IHC (P)

          Immunohistochemistry was performed on cancer biopsies of deparaffinized Human cervical carcinoma tissue. To expose target proteins, heat induced antigen retrieval was performed using 10mM sodium citrate (pH6.0) buffer, microwaved for 8-15 minutes. Following antigen retrieval tissues were blocked in 3% BSA-PBS for 30 minutes at room temperature. Tissues were then probed at a dilution of 1:20 with a rabbit po... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PMCA2 ATPase Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PMCA2 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA2 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA2 ATPase Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Immunohistochemistry (IHC)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody in Western Blot (WB)
          PMCA2 ATPase Antibody
          PMCA2 ATPase Antibody
          PMCA2 ATPase Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PMCA2 ATPase Polyclonal Antibody

          Product Details

          PA1-915

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1:20-1:200
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 12 publications 12 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Gel Shift (EMSA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Pig, Rat

          Published species

          Chicken, Hamster, Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic Peptide: T(5) N S D F Y S K N Q R N E S S(19)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2243199

          Product Specific Information

          PA1-915 detects calcium pump of the plasma membrane 2 (PMCA2 ATPase) from human and rat and porcine tissues and cells.

          PA1-915 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~127 kDa protein and an ~133 kDa protein representing PMCA2a and PMCA2b ATPase, respectively, from rat brain microsomal fractions.

          PA1-915 immunizing peptide corresponds to amino acid residues 5-19 from human PMCA2 ATPase protein. This sequence is completely conserved between human and rat PMCA2 ATPase.

          Target Information

          The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase, Ca++ transporting, plasma membrane 2; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane calcium ATPase; plasma membrane calcium pump; plasma membrane calcium-ATPase isoform 2 alternative splice variant a at splice site C; plasma membrane calcium-ATPase isoform 2, splice variant i

          View more View less

          Gene Aliases: DFNA82; PMCA2; PMCA2a; PMCA2i

          View more View less

          UniProt ID: (Human) Q01814, (Rat) P11506

          View more View less

          Entrez Gene ID: (Human) 491, (Pig) 100625418, (Rat) 24215

          View more View less

          Function(s)
          nucleotide binding calcium-transporting ATPase activity calcium ion binding protein binding calmodulin binding ATP binding ATPase activity PDZ domain binding calcium-dependent ATPase activity P-type calcium transporter activity involved in regulation of postsynaptic cytosolic calcium ion concentration glutamate receptor binding primary active transporter
          Process(es)
          calcium ion transport sensory perception of sound neuron differentiation ion transmembrane transport regulation of cytosolic calcium ion concentration calcium ion transmembrane transport cochlea development regulation of postsynaptic cytosolic calcium ion concentration regulation of cardiac conduction cell morphogenesis neural retina development cellular calcium ion homeostasis lactation locomotory behavior regulation of cell size cerebellum development cerebellar Purkinje cell layer morphogenesis cerebellar Purkinje cell differentiation cerebellar granule cell differentiation locomotion serotonin metabolic process inner ear morphogenesis otolith mineralization cGMP metabolic process regulation of synaptic plasticity inner ear development synapse organization neuromuscular process controlling balance detection of mechanical stimulus involved in sensory perception of sound positive regulation of calcium ion transport auditory receptor cell stereocilium organization inner ear receptor cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch