Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ERP29 Antibodies

          Invitrogen

          ERP29 Polyclonal Antibody

          3 References
          View all (15) ERP29 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ERP29 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ERP29 Antibody (PA3-011) in ICC/IF

          Immunofluorescent analysis of ERp29 using anti-ERp29 polyclonal antibody (Product # PA3-011) shows staining in NS-1 Cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          ERP29 Antibody in Immunocytochemistry (ICC/IF)
          ERP29 Antibody in Western Blot (WB)
          ERP29 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ERP29 Polyclonal Antibody

          Product Details

          PA3-011

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:500
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          1:500
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Guinea pig, Hamster, Human, Mouse, Non-human primate, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          6-histidine epitope tagged full length recombinant rat ERp29.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2101510

          Product Specific Information

          PA3-011 detects ERp29 protein in rat, mouse, hamster, canine, human, non-human primate, bovine and guinea pig samples.

          PA3-011 has successfully been used in immunofluorescence, immunoprecipitation and Western blot procedures. By Western blot, this antibody detects an ~29 kDa protein representing ERp29 in rat liver homogenate. Immunofluorescence data demonstrates that ERp29 is localized to the ER of rat thyrocytes using PA3-011.

          The PA3-011 immunogen was a 6-histidine epitope tagged full length recombinant ERp29 protein.

          Target Information

          This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C12orf8; endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein 29; Endoplasmic reticulum protein ERp29; endoplasmic reticulum resident protein 28; endoplasmic reticulum resident protein 29; endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; ERp31; protein disulfide isomerase family A, member 9; unnamed protein product

          View more View less

          Gene Aliases: 1200015M03Rik; 2810446M09Rik; C12orf8; ERp28; ERp31; HEL-S-107; PDI-DB; PDIA9

          View more View less

          UniProt ID: (Bovine) P81623, (Human) P30040, (Mouse) P57759, (Rat) P52555

          View more View less

          Entrez Gene ID: (Bovine) 613357, (Dog) 477482, (Human) 10961, (Chinese hamster) 100762501, (Guinea pig) 100731699, (Mouse) 67397, (Rat) 117030

          View more View less

          Function(s)
          ATP binding ATPase activity protein disulfide isomerase activity protein binding protein homodimerization activity chaperone binding membrane traffic protein
          Process(es)
          activation of MAPK activity protein secretion positive regulation of gene expression negative regulation of gene expression negative regulation of protein secretion regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway protein folding intracellular protein transport ER to Golgi vesicle-mediated transport response to hormone protein unfolding positive regulation of MAPK cascade response to endoplasmic reticulum stress
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0