Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cyp4a10 Antibodies

          Invitrogen

          Cyp4a10 Polyclonal Antibody

          4 Published Figures
          13 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cyp4a10 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (4)
          Cyp4a10 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cyp4a10 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cyp4a10 Antibody (PA3-033) in ICC/IF

          Immunofluorescent analysis of Cytochrome P450 4A (green) showing staining in the cytoplasm and membrane of H-4-II-E cells. Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a Cytochrome P450 4A polyclonal antibody (Product # PA3-033) in 3% BSA-PBS at a dilution of 1:100 and incubated overnight at 4 °C in a humidified chamber. Cells were washed with PBST and incubated with a DyLight-conjugated secondary antibody in PBS... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cyp4a10 Antibody in Immunocytochemistry (ICC/IF)
          Cyp4a10 Antibody in Immunocytochemistry (ICC/IF)
          Cyp4a10 Antibody in Immunocytochemistry (ICC/IF)
          Cyp4a10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cyp4a10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cyp4a10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cyp4a10 Antibody in Western Blot (WB)
          Cyp4a10 Antibody in Western Blot (WB)
          Cyp4a10 Antibody in Western Blot (WB)
          Cyp4a10 Antibody in Western Blot (WB)
          Cyp4a10 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cyp4a10 Polyclonal Antibody

          Product Details

          PA3-033

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:2,000
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Cat, Human, Rabbit, Rat

          Published species

          Hamster, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues D(431) P S R F A P D S P R H S H S(445) of rat Cytochrome P450 4A.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum, PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325980

          Product Specific Information

          PA3-033 detects cytochrome P450 4A (isoforms 1, 2, & 3) from human, feline, rabbit and rat tissues.

          PA3-033 has been successfully used in Western blot, ICC/IF and IHC procedures. By Western blot, this antibody detects an ~50 kDa protein representing P450 4A in rat liver extract. Immunohistochemical staining of P450 4A in rat brain with PA3-033 results in perivascular and surrounding cortex staining.

          The PA3-033 immunogen is a synthetic peptide corresponding to residues D(431) P S R F A P D S P R H S H S(445) of rat Cytochrome P450 4A.

          Target Information

          The cytochrome P450 (P450) family of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as NADPH and P450 reductase. There are over 200 cDNA's which encode P450 protein. Epoxygenases are those P450 proteins which metabolize AA to epoxyeicosatrienoic acid (EETs) and w-hydroxylases are those P450 proteins which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). 20-HETE is converted from AA by the 4A family of P450 proteins which includes at least 8 different, though closely related isoforms. 4A1, 4A2, & 4A3 have been cloned from liver, kidney and testis and have been detected in renal, hepatic & brain microvessels.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CYP; cytochrome P450, 4A1; cytochrome P450, 4a10; cytochrome P450, family 4, subfamily a, polypeptide 10; cytochrome P450, family 4, subfamily A, polypeptide 22; cytochrome P450, IVA1; cytochrome P450-LA-omega 1; cytochrome P452; lauric acid omega-hydroxylase; long-chain fatty acid omega-monooxygenase; P450

          View more View less

          Gene Aliases: Cyp4a10; Cyp4a22; CYPIVA10

          View more View less

          UniProt ID: (Rat) P08516

          View more View less

          Entrez Gene ID: (Rat) 50549

          View more View less

          Function(s)
          monooxygenase activity iron ion binding arachidonic acid monooxygenase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen alkane 1-monooxygenase activity heme binding arachidonic acid omega-hydroxylase activity long-chain fatty acid omega-hydroxylase activity 16-hydroxypalmitate dehydrogenase activity long-chain fatty acid omega-1 hydroxylase activity fatty acid omega-1 hydroxylase activity medium-chain fatty acid omega-hydroxylase activity
          Process(es)
          kidney development placenta development lipid hydroxylation arachidonic acid metabolic process linoleic acid metabolic process icosanoid biosynthetic process lauric acid metabolic process omega-hydroxylase P450 pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch